Skip to Content
Merck
All Photos(1)

Key Documents

M-132

Supelco

MSTFA Reagent

ampule of 10 × 1.2 mL, analytical standard, Cerilliant®

Synonym(s):

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, N-Trimethylsilyl-N-methyl trifluoroacetamide, MSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CON(CH3)Si(CH3)3
CAS Number:
Molecular Weight:
199.25
Beilstein:
1941550
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

8.8 mmHg ( 27 °C)

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

packaging

ampule of 10 × 1.2 mL

manufacturer/tradename

Cerilliant®

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.38 (lit.)

bp

130-132 °C (lit.)

density

1.075 g/mL at 25 °C (lit.)

application(s)

forensics and toxicology

format

neat

storage temp.

−20°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C

InChI

1S/C6H12F3NOSi/c1-10(12(2,3)4)5(11)6(7,8)9/h1-4H3

InChI key

MSPCIZMDDUQPGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MSTFA (N-Methyl-N-trimethylsilylfluoroacetamide) is an important trimethylsilyl reagent, and a commonly applied reagent since it is highly reactive towards a broad range of compound classes. GC-MS optimization studies reveal the use of this reagent either alone or in combination with 1% of trimethylchlorosilane as a catalyst for silylation of complex mixtures. It has similar reactivity as BSA and BSTFA.

Application

MSTFA reagent may be used as a derivatizing agent in the following:
  • Analysis of estrogen steroids such as estrone (E1), 17α-ethinylestradiol (EE2), and trenbolone acetate metabolites, melengestrol in environmental matrices using gas chromatography coupled to mass spectrometry (GC-MS).
  • Quantification of ibuprofen, naproxen, ketoprofen, and diclofenac in wastewater samples using gas chromatography coupled to mass spectrometry (GC-MS).

Features and Benefits

  • Very volatile of TMS-acetamides.
  • More volatile than BSA or BSTFA but with similar silylation strength.
  • Useful in the analysis of volatile trace materials.
  • Used in the preparation of volatile and thermally stable derivatives for GC and MS analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of trimethylsilyl cyanide derivatization for robust and broad-spectrum high-throughput gas chromatography?mass spectrometry based metabolomics
Khakimov B, et al.
Analytical and Bioanalytical Chemistry, 405(3), 9193-9205 (2013)
Identification and quantification of ibuprofen, naproxen, ketoprofen and diclofenac present in waste-waters, as their trimethylsilyl derivatives, by gas chromatography mass spectrometry
Sebok A, et al.
Talanta, 76(3), 642-650 (2008)
Analysis of trenbolone acetate metabolites and melengestrol in environmental matrices using gas chromatography--tandem mass spectrometry
Parker AJ, et al.
Talanta, 99(3), 238-246 (2012)
Sumandeep Rana et al.
Journal of analytical toxicology, 32(5), 355-363 (2008-06-12)
A method for the simultaneous determination of six commonly prescribed cyclic antidepressants and their major metabolites in urine is presented. This method can be used for quantitation of amitriptyline, nortriptyline, imipramine, desipramine, doxepin, desmethyldoxepin, and maprotiline in human urine, in
Harin Kanani et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 871(2), 191-201 (2008-06-10)
Metabolomics being the most recently introduced "omic" analytical platform is currently at its development phase. For the metabolomics to be broadly deployed to biological and clinical research and practice, issues regarding data validation and reproducibility need to be resolved. Gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service