Merck
  • Trace determination of perchlorate using electromembrane extraction and capillary electrophoresis with capacitively coupled contactless conductivity detection.
Trace determination of perchlorate using electromembrane extraction and capillary electrophoresis with capacitively coupled contactless conductivity detection.
Electrophoresis (2011-10-18)
Isaac K Kiplagat, Thi Kieu Oanh Doan, Pavel Kubáň, Petr Boček
ABSTRACT
Electromembrane extraction (EME) and CE with capacitively coupled contactless conductivity detection (CE-C(4) D) was applied to rapid and sensitive determination of perchlorate in drinking water and environmental samples. Porous polypropylene hollow fiber impregnated with 1-heptanol acted as a supported liquid membrane (SLM) and perchlorate was transported and preconcentrated in the fiber lumen on application of electric field. High selectivity of perchlorate determination and its baseline separation from major inorganic anions was achieved in CE-C(4) D using background electrolyte solution consisting of 7.5 mM L-histidine and 40 mM acetic acid at pH 4.1. The analytical method showed excellent parameters in terms of reproducibility; RSD values for migration times and peak areas at a spiked concentration of 15 μg/L of perchlorate (US EPA recommended limit for drinking water) were below 0.2 and 8.7%, respectively, in all examined water samples. Linear calibration curves were obtained for perchlorate in the concentration range 1-100 μg/L (r(2) ≥0.999) with limits of detection at 1 μg/L for tap water and at 0.25-0.35 μg/L for environmental and bottled potable water samples. Recoveries at 15 μg/L of perchlorate were between 95.9 and 106.7% with minimum and maximum recovery values for snow and bottled potable water samples, respectively.
Sigma-Aldrich
1-Heptanol, ≥99.5% (GC)
Sigma-Aldrich
1-Heptanol, 98%
Supelco
1-Heptanol, analytical standard, ≥99.5% (GC)
Sigma-Aldrich
Heptyl alcohol, ≥97%, FCC, FG