Merck
  • PdCl2-catalyzed efficient allylation and benzylation of heteroarenes under ligand, base/acid, and additive-free conditions.
PdCl2-catalyzed efficient allylation and benzylation of heteroarenes under ligand, base/acid, and additive-free conditions.
Chemical communications (Cambridge, England) (2011-04-01)
Feng-Quan Yuan, Lian-Xun Gao, Fu-She Han
ABSTRACT
We present a PdCl(2)-catalyzed protocol for highly efficient allylation and benzylation of a rich variety of N-, O-, and S-containing heteroarenes under base/acid, additive, and ligand-free conditions. The method represents the very few examples for simple, universally applicable, clean, and atom-efficient functionalization of heteroarenes.
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
Palladium(II) chloride ChemBeads
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis