Merck
  • A novel multifunctional pharmaceutical excipient: modification of the permeability of starch by processing with magnesium silicate.
A novel multifunctional pharmaceutical excipient: modification of the permeability of starch by processing with magnesium silicate.
International journal of pharmaceutics (2011-03-23)
Iyad Rashid, Mayyas Al-Remawi, Stephen A Leharne, Babur Z Chowdhry, Adnan Badwan
ABSTRACT
A directly compressible excipient has been developed by co-processing starch with magnesium silicate. The foregoing was achieved either by co-precipitation of magnesium silicate onto different types of starch or by dry granulation of maize starch with magnesium silicate. A variety of techniques (permeability, water retention/swelling, compression analysis, scanning electron microscopy, tensile strength and disintegration/dissolution studies) were used to characterize these systems. The permeability of the formulations produced using the two methods was evaluated experimentally using Darcy's permeability law. Magnesium silicate, as an anti-adhering agent, increases the permeability of both maize and partially pregelatinized starch, resulting in compacts of high mechanical strength, short disintegration time and low lubricant sensitivity. Such advantages are evident when the properties of the physical mixture of maize starch with magnesium silicate are compared with the co-precipitation and dry granulation techniques. Formulation with this novel excipient system, using paracetamol as a model drug, indicated its suitability as a single multifunctional excipient.
Product Description
Supelco
Florisil® Adsorbent for Chromatography, 60-100 mesh
Supelco
Florisil® Adsorbent for Chromatography, 30-60 mesh
Supelco
Florisil®, 100-200 mesh, fine powder
Supelco
Florisil®, for the determination of hydrocarbon acc. to ISO 9377-2
Supelco
Florisil®, <200 mesh, fine powder
Supelco
Florisil® PR
Supelco
Florisil®, 60-100 mesh, coarse powder
Sigma-Aldrich
Florisil®, 100-200 mesh