Skip to Content
Merck
All Photos(1)

Key Documents

20280-U

Supelco

Florisil® Inorganic Sorbent

PR grade, 60-100 mesh, coarse powder

Synonym(s):

Activated magnesium silicate, Magnesium silicate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

product name

Florisil®, PR grade, 60-100 mesh, coarse powder

grade

PR grade

Quality Level

form

coarse powder

packaging

jar of 454 g

technique(s)

LPLC: suitable

surface area

289 m2/g

matrix

Magnesium silicate hydrate

matrix active group

magnesium silicate hydrate

particle size

149-250 μm
60-100 mesh

pore size

60 Å mean pore size

suitability

suitable for (Changes of Official Methods of Analysis, JAOAC, Chapter 24, 208(h), Vol. 49, p 233, 1966.)

separation technique

hydrophilic interaction (HILIC)

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Florisil, adsorbent for the separation of lipids is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.

Application

  • Florisil was used as column packing material in column chromatography.
  • Florisil was used as an adsorbent in gas chromatography with electron capture detector (GC/ECD), capillary column chromatography and adsorption column chromatography.
  • Florisil was used in clean-up procedures in gas chromatography-tandem mass spectrometry and GC-FID.

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved gas chromatographic method for the determination of perfluorocarbon tracers in the atmosphere
Lagomarsino, Raymond J.
Journal of Chromatographic Science, 34, 405-412 (1996)
Andrew C Singer et al.
Environmental toxicology and chemistry, 22(9), 1998-2004 (2003-09-10)
This study investigated the interactive effects of bioaugmentation, biostimulation, and the rhizosphere during remediation of Aroclor 1242-contaminated soil. Treatments were repeatedly augmented with polychlorinated bipheny (PCB)-degrading bacteria, inducers (carvone and salicylic acid), surfactant (sorbitan trioleate), minimal salts medium in a
R C Prados-Rosales et al.
Journal of chromatography. A, 993(1-2), 121-129 (2003-05-09)
A rapid analytical method for determination of organochlorine pesticide residues in sunflower seeds based on focused microwave-assisted Soxhlet extraction has been developed. The main factors affecting the extraction efficiency--namely microwave power, irradiation time, volume of extractant and number of cycles--were
Determination of atmospheric perfluorocarbon background concentrations of fL/L range at the western coastal area of Korea
Kim, HyeKyeong, et al.
Atmospheric Environment, 1, 4-4 (2002)
The Florisil? catalyzed [1, 3]-sigmatropic shift of allyl phenyl ethers-An entryway into novel mycophenolic acid analogues.
Talamas, Francisco X., et al.
Tetrahedron Letters, 38, 4725-4728 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service