Merck
  • Inhibitory effects of citronellol and geraniol on nitric oxide and prostaglandin E₂production in macrophages.
Inhibitory effects of citronellol and geraniol on nitric oxide and prostaglandin E₂production in macrophages.
Planta medica (2010-05-28)
Yu-Wen Su, Shiou-Huei Chao, Meng-Hwan Lee, Tsang-Yow Ou, Ying-Chieh Tsai
ABSTRACT
Geranium oil has been used traditionally for diarrhea, dermatitis, and intestinal inflammation in East Asia. The aim of this study was to determine the effects of geranium oil's characteristic components, citronellol and geraniol, on lipopolysaccharide (LPS)-induced nitric oxide (NO) and prostaglandin E(2) (PGE(2)) production in RAW 264.7 macrophages. Citronellol and geraniol suppressed NO and PGE(2) production in a dose-dependent manner. The inhibitory efficacy of geraniol was concomitant with decreases in protein and mRNA expression levels of inducible nitric oxide synthase (iNOS), whereas citronellol inhibited only iNOS enzymatic activity. By adding citronellol and geraniol, the LPS-induced cyclooxygenase-2 (COX-2) protein and mRNA expression levels were significantly attenuated, whereas cytosolic degradation of I κB α and upregulation of NF-κB p65 in the nucleus were reversed. These results suggested that citronellol and geraniol exhibit anti-inflammatory activities, supporting their common use and demonstrating their therapeutic potential for inflammation-associated disorders.
Sigma-Aldrich
(R)-(+)-β-Citronellol, 95%
Sigma-Aldrich
(S)-(−)-β-Citronellol, ≥97%
Supelco
(−)-β-Citronellol, analytical standard
Supelco
(±)-β-Citronellol, analytical standard
Sigma-Aldrich
Citronellol, mixture of isomers, natural, ≥95%, FG
Sigma-Aldrich
β-Citronellol, 95%
Sigma-Aldrich
Citronellol, ≥95%, FCC, FG