Merck
  • Efficient production of 2-oxobutyrate from 2-hydroxybutyrate by using whole cells of Pseudomonas stutzeri strain SDM.
Efficient production of 2-oxobutyrate from 2-hydroxybutyrate by using whole cells of Pseudomonas stutzeri strain SDM.
Applied and environmental microbiology (2010-01-19)
Chao Gao, Wen Zhang, Chuanjuan Lv, Lixiang Li, Cuiqing Ma, Chunhui Hu, Ping Xu
ABSTRACT
2-Oxobutyrate is an important intermediate in the chemical, drug, and food industries. Whole cells of Pseudomonas stutzeri SDM, containing NAD-independent lactate dehydrogenases, effectively converted 2-hydroxybutyrate into 2-oxobutyrate. Under optimal conditions, the biocatalytic process produced 2-oxobutyrate at a high concentration (44.4 g liter(-1)) and a high yield (91.5%).
Sigma-Aldrich
2-Ketobutyric acid, 97%
Sigma-Aldrich
2-Oxobutyric acid, ≥95%, FG
Sigma-Aldrich
Sodium 2-oxobutyrate, powder