Merck
  • Nondestructive, colorimetric monitoring of amines and thiols on a solid support.
Nondestructive, colorimetric monitoring of amines and thiols on a solid support.
Organic letters (2009-07-09)
Seh-Jin Yang, Xi Zhe Tian, Injae Shin
ABSTRACT
A new, nondestructive, highly sensitive method for colorimetric monitoring of primary amines, secondary amines, and thiols on a solid support was developed. The resin used in this method is simply regenerated for the repetition of the reaction or an ensuing reaction. By using this new method, several peptides containing secondary amide linkages and C-terminal hydrazide groups were prepared in high purities and yields.
Sigma-Aldrich
4-Ethylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
4-Ethylmorpholine, ≥97.0% (GC)
Sigma-Aldrich
4-Ethylmorpholine, 97%
Sigma-Aldrich
Disperse Red 1, Dye content 95 %