Skip to Content
Merck
All Photos(2)

Key Documents

344206

Sigma-Aldrich

Disperse Red 1

Dye content 95 %

Synonym(s):

N-Ethyl-N-(2-hydroxyethyl)-4-(4-nitrophenylazo)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N4O3
CAS Number:
Molecular Weight:
314.34
Colour Index Number:
11110
Beilstein:
5353614
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

composition

Dye content, 95%

mp

160-162 °C (lit.)

λmax

502 nm

SMILES string

CCN(CCO)c1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C16H18N4O3/c1-2-19(11-12-21)15-7-3-13(4-8-15)17-18-14-5-9-16(10-6-14)20(22)23/h3-10,21H,2,11-12H2,1H3/b18-17+

InChI key

FOQABOMYTOFLPZ-ISLYRVAYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Disperse Red 1 (DR1) is a non-linear optical (NLO) material which is a chromophore used as an azobenzene dye. It can form a dipole network that can enhance the electro-optic effect of the non-linear components and can also improve the photorefractive effect.

Application

DR1 dye can be incorporated into hydroxypropyl cellulose (HPC) matrix which can be used as a coating that finds potential application in light storage and photonics. It can also be used to evaluate the third order non-linear susceptibility (X(3)) for 1,3-indandione based glasses for use in optical frequency conversion. Polytetrafluoroethylene (PTFE) membranes modified using DR1 maintain the mass transport and diffusion properties under direct light irradiation and can be used for water desalination applications.

Features and Benefits

This benchmark dipolar chromophore was incorporated into aromatic polymers to obtain model electrooptic polymer films.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A calix (4) resorcinarene-based hyper-structured molecule bearing disperse red 1 as the chromophore with enhanced photorefractive performance under non-electric field.
Zong W, et al.
Dyes and Pigments, 160(10), 579-586 (2019)
Water desalination using visible light by disperse red 1 modified PTFE membrane.
Fujiwara M.
Desalination, 404(1), 79-86 (2017)
Reversible light-induced solubility of disperse red 1 dye in a hydroxypropyl cellulose matrix.
Pawlicka A, et al.
Cellulose, 25(3), 2083-2090 (2018)
Arylmethylene-1, 3-indandione based molecular glasses: Third order optical non-linearity.
Seniutinas G, et al.
Dyes and Pigments, 95(1), 33-40 (2012)
Polymeric electrooptic composite based on Disperse Red and its derivatives for use in photonics
Burunkova YE, et al.
Journal of Optical Technology, 77(10), 643-648 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service