Merck
  • Neuroprotection by a selective estrogen receptor beta agonist in a mouse model of global ischemia.
Neuroprotection by a selective estrogen receptor beta agonist in a mouse model of global ischemia.
American journal of physiology. Heart and circulatory physiology (2004-05-25)
H V O Carswell, I M Macrae, L Gallagher, E Harrop, K J Horsburgh
ABSTRACT
The present study employs selective estrogen receptor (ER) agonists to determine whether 17beta-estradiol-induced neuroprotection in global ischemia is receptor mediated and, if so, which subtype of receptor (ERalpha or ERbeta) is predominantly responsible. Halothane-anesthetized female C57Bl/6J mice were ovariectomized, and osmotic minipumps containing ERbeta agonist diarylpropiolnitrile (DPN) (8 mg.kg(-1).day(-1), n = 12) or vehicle (50% DMSO in 0.9% saline) (n = 9) or ERalpha agonist propyl pyrazole triol (PPT) (2 mg.kg(-1).day(-1), n = 13) or vehicle (50% DMSO in 0.9% saline) (n = 10) were implanted subcutaneously. One week later transient global ischemia was induced by bilateral carotid artery occlusion under halothane anesthesia, and the mice were perfusion fixed 72 h later. ERbeta agonist DPN significantly reduced ischemic damage by 70% in the caudate nucleus and 55% in the CA1 region compared with vehicle controls (P < 0.05, Mann-Whitney U-statistic). In contrast, pretreatment with the ERalpha agonist PPT had no effect on the extent of neuronal damage compared with controls. The data indicate a significant estrogen receptor-mediated neuroprotection in a global cerebral ischemia model involving ERbeta.
Product Description
Supelco
Propionitrile, analytical standard
Sigma-Aldrich
Propionitrile, 99%
Sigma-Aldrich
Propionitrile, purum, ≥99.0% (GC)