Merck
  • Functional and anionic cellulose-interacting polymers by selective chemo-enzymatic carboxylation of galactose-containing polysaccharides.
Functional and anionic cellulose-interacting polymers by selective chemo-enzymatic carboxylation of galactose-containing polysaccharides.
Biomacromolecules (2012-06-26)
Kirsti Parikka, Ann-Sofie Leppänen, Chunlin Xu, Leena Pitkänen, Paula Eronen, Monika Osterberg, Harry Brumer, Stefan Willför, Maija Tenkanen
ABSTRACT
Carboxylated, anionic polysaccharides were selectively prepared using a combination of enzymatic and chemical reactions. The galactose-containing polysaccharides studied were spruce galactoglucomannan, guar galactomannan, and tamarind galactoxyloglucan. The galactosyl units of the polysaccharides were first oxidized with galactose oxidase (EC 1.1.3.9) and then selectively carboxylated, resulting in the galacturonic acid derivatives with good conversion and yield. The degrees of oxidation (DO) of the products were determined by gas chromatography-mass spectrometry (GC-MS). A novel feasible electrospray ionization-mass spectrometry (ESI-MS) method was also developed for the determination of DO. The solution properties and charge densities of the products were investigated. The interaction of the products with cellulose was studied by two methods, bulk sorption onto bleached birch kraft pulp and adsorption onto nanocellulose ultrathin films by quartz crystal microbalance with dissipation (QCM-D). To study the effect of the location of the carboxylic acid groups on the physicochemical properties, polysaccharides were also oxidized by 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO)-mediated reaction producing polyuronic acids. The chemo-enzymatically oxidized galacturonic polysaccharides with an unmodified backbone had a better ability to interact with cellulose than the TEMPO-oxidized products. The selectively carboxylated polysaccharides can be further exploited, as such, or in the targeted functionalization of cellulose surfaces.
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Galactose Oxidase from Dactylium dendroides, ≥30 units/mg solid
Sigma-Aldrich
Guar
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Galactose Oxidase from Dactylium dendroides, lyophilized powder, ≥3,000 units/g solid
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Gold etchant, standard