Merck
  • Enzymatic primer-extension with glycerol-nucleoside triphosphates on DNA templates.
Enzymatic primer-extension with glycerol-nucleoside triphosphates on DNA templates.
PloS one (2009-03-24)
Jesse J Chen, Ching-Hsuan Tsai, Xin Cai, Allen T Horhota, Larry W McLaughlin, Jack W Szostak
ABSTRACT
Glycerol nucleic acid (GNA) has an acyclic phosphoglycerol backbone repeat-unit, but forms stable duplexes based on Watson-Crick base-pairing. Because of its structural simplicity, GNA is of particular interest with respect to the possibility of evolving functional polymers by in vitro selection. Template-dependent GNA synthesis is essential to any GNA-based selection system. In this study, we investigated the ability of various DNA polymerases to use glycerol-nucleoside triphosphates (gNTPs) as substrates for GNA synthesis on DNA templates. Therminator DNA polymerase catalyzes quantitative primer-extension by the incorporation of two glyceronucleotides, with much less efficient extension up to five glyceronucleotides. Steady-state kinetic experiments suggested that GNA synthesis by Therminator was affected by both decreased catalytic rates and weakened substrate binding, especially for pyrimidines. In an attempt to improve pyrimidine incorporation by providing additional stacking interactions, we synthesized two new gNTP analogs with 5-propynyl substituted pyrimidine nucleobases. This led to more efficient incorporation of gC, but not gT. We suggest that directed evolution of Therminator might lead to mutants with improved substrate binding and catalytic efficiency.
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
2-Chloro-1,3,2-benzodioxaphosphorin-4-one, 95%
Sigma-Aldrich
5-Iodocytosine
Sigma-Aldrich
Ammonium citrate tribasic, ≥97% (titration)
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tris(tetrabutylammonium) hydrogen pyrophosphate, ≥97.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium sulfite, ≥98%
Sigma-Aldrich
5-Iodouracil, 98%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Sodium sulfite, anhydrous, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Supelco
3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
DEAE–Sephadex® A-25 chloride form
Supelco
Silica gel, high-purity grade (9385), pore size 60 Å, 230-400 mesh particle size
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Triethylammonium bicarbonate buffer, 1.0 M, pH 8.5±0.1
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Tetrakis(triphenylphosphine)palladium(0), 99%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%