Skip to Content
Merck
All Photos(3)

Key Documents

93397

Sigma-Aldrich

Tris(tetrabutylammonium) hydrogen pyrophosphate

≥97.0% (calc. on dry substance, T)

Synonym(s):

Pyrophosphoric acid tris(tetrabutylammonium) salt, Tetrabutylammonium pyrophosphate (3:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3CH2CH2CH2)4N+]3[O=P(O-)2OP(=O)(O-)OH]
CAS Number:
Molecular Weight:
902.34
Beilstein:
4632762
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (calc. on dry substance, T)

form

crystals

impurities

≤8% water

storage temp.

2-8°C

SMILES string

OP([O-])(=O)OP([O-])([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/3C16H36N.H4O7P2/c3*1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-8(2,3)7-9(4,5)6/h3*5-16H2,1-4H3;(H2,1,2,3)(H2,4,5,6)/q3*+1;/p-3

InChI key

BMTUFQKYWWLCLC-UHFFFAOYSA-K

Application

Other Notes

Reagent for pyrophosphorylation; reaction of isoprenoid derivatives; Pyrophosphorylation of nucleosides; Triphosphorylation of nucleosides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of allylic and homoallylic isoprenoid pyrophosphates.
V J Davisson et al.
Methods in enzymology, 110, 130-144 (1985-01-01)
A.B. Woodside et al.
Organic Syntheses, 66, 211-211 (1988)
The Journal of Organic Chemistry, 51, 4768-4768 (1986)
V.J. Davisson et al.
The Journal of Organic Chemistry, 52, 1794-1794 (1987)
F. Seela et al.
Helvetica Chimica Acta, 74, 554-554 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service