Merck
  • A Comparison of the α2/3/5 Selective Positive Allosteric Modulators L-838,417 and TPA023 in Preclinical Models of Inflammatory and Neuropathic Pain.
A Comparison of the α2/3/5 Selective Positive Allosteric Modulators L-838,417 and TPA023 in Preclinical Models of Inflammatory and Neuropathic Pain.
Advances in pharmacological sciences (2011-12-14)
Sarah Nickolls, Hannah Mace, Rebecca Fish, Michelle Edye, Rachel Gurrell, Magnus Ivarsson, Tom Pitcher, Sachi Tanimoto-Mori, Denise Richardson, Catherine Sweatman, Janet Nicholson, Cameron Ward, John Jinks, Christine Bell, Kimberly Young, Huw Rees, Andrew Moss, Ross Kinloch, Gordon McMurray
ABSTRACT
GABA(A) receptors containing α2/3 subunits are current targets in the battle to develop new pain medications, as they are expressed in the spinal cord where increasing inhibitory drive should result in analgesia. However, this approach is prone to a range of side effects including sedation, cognitive impairment, and abuse as a consequence of the widespread influence of GABA. The ability to make subtype selective low-efficacy benzodiazepine compounds, which potentiate the action of GABA at specific α subunits, has the potential to reduce this side effect profile. In this study, we have investigated the effects of the medium-efficacy positive allosteric modulator (PAM) L-838,417 and the low-efficacy PAM TPA023 in a number of preclinical inflammatory and neuropathic pain models. We conclude that either the higher level of efficacy at α2/3 or efficacy at α5 is required for compounds to have a significant analgesic effect in a range of models, and, therefore, although the side-effect profile of compounds can be reduced compared to typical benzodiazepines, it is unlikely that it can be completely eliminated.
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Polyethylene glycol/ dimethyl sulfoxide solution, Hybri-Max, average mol wt 1,450, 50 % (w/v), 0.2 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (methoxyl group, on Dry Basis), meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Bretazenil, ≥96% (HPLC), solid
Sigma-Aldrich
Polyethylene glycol solution, BioUltra, for molecular biology, 1,000, ~50% in H2O
Sigma-Aldrich
Glycerol formal, ≥98.0% (GC)
Sigma-Aldrich
Glycerol formal, 47-67% 5-hydroxy-1,3-dioxane basis (GC), 33-53% 4-hydroxymethyl-1,3-dioxolane basis (GC)
Sigma-Aldrich
Polyethylene glycol solution, Hybri-Max, 50 % (w/v), average mol wt 1,450, 0.2 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Polyethylene glycol solution, 40 % (w/w) in H2O, average mol wt 8,000