Skip to Content
Merck
All Photos(1)

Key Documents

64632

Sigma-Aldrich

Methyl cellulose

tested according to Ph. Eur.

Synonym(s):

Methylcellulosum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

powder

color

white to off-white

useful pH range

5.0-8.0

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Paolo Vitiello et al.
Journal of experimental & clinical cancer research : CR, 40(1), 15-15 (2021-01-08)
Despite the advancements in new therapies for colorectal cancer (CRC), chemotherapy still constitutes the mainstay of the medical treatment. For this reason, new strategies to increase the efficacy of chemotherapy are desirable. Poly-ADP-Ribose Polymerase inhibitors (PARPi) have shown to increase
Yihua Wang et al.
Biochemistry, 52(9), 1611-1621 (2013-02-07)
Myosin powers contraction in heart and skeletal muscle and is a leading target for mutations implicated in inheritable muscle diseases. During contraction, myosin transduces ATP free energy into the work of muscle shortening against resisting force. Muscle shortening involves relative
Walter Fuchs et al.
Journal of virology, 83(8), 3930-3943 (2009-02-06)
Cleavage and encapsidation of newly replicated herpes simplex virus type 1 (HSV-1) DNA requires several essential viral gene products that are conserved in sequence within the Herpesviridae. However, conservation of function has not been analyzed in greater detail. For functional
Hisham Al-Obaidi et al.
International journal of pharmaceutics, 443(1-2), 95-102 (2013-01-10)
Solid dispersions of varying weight ratios compositions of the nonionic drug, griseofulvin and the hydrophilic, anionic polymer, hydroxylpropylmethyl cellulose acetate succinate, have been prepared by ball milling and the resulting samples characterized using a combination of Fourier transform infra-red spectroscopy
Elsa-Noah N'Diaye et al.
EMBO reports, 10(2), 173-179 (2009-01-17)
Ubiquilins (UBQLNs) are adaptor proteins thought to deliver ubiquitinated substrates to proteasomes. Here, we show a role for UBQLN in autophagy: enforced expression of UBQLN protects cells from starvation-induced death, whereas depletion of UBQLN renders cells more susceptible. The UBQLN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service