Merck
  • Human dendritic cells infected with an adenoviral vector suppress proliferation of autologous and allogeneic T cells.
Human dendritic cells infected with an adenoviral vector suppress proliferation of autologous and allogeneic T cells.
Immunology (2008-05-31)
Katy R Newton, Emma Sala-Soriano, Hemlata Varsani, John R Stephenson, David Goldblatt, Lucy R Wedderburn
ABSTRACT
Dendritic cells (DCs) play a key role in the type and course of an immune response. The manipulation of human DCs to produce therapeutic agents by transduction with viral vectors is a growing area of research. We present an investigation into the effects of adenoviral vector infection on human DCs and other cell types, and on their subsequent ability to induce T-cell proliferation. We show that infection with replication-deficient adenovirus results in impaired proliferation of T cells in a mixed lymphocyte reaction (MLR). We show this to be an active suppression rather than a defect in the DCs as T cells also fail to proliferate in response to phytohaemagglutinin in the presence of adenoviral vector-infected DCs. This suppression is not attributable to phenotypic changes, death or inability of the DCs to produce cytokines on stimulation. By separation of DCs from T cells, and addition of conditioned supernatants, we show that suppression is mediated by a soluble factor. Blocking of interleukin (IL)-10 but not transforming growth factor (TGF)-beta could overcome the suppressive effect in some donors, and the source of the suppressive IL-10 was lymphocytes exposed to conditioned supernatant. Together our data suggest that infection of DCs by adenoviral vectors leads to suppression of the resulting immune response.
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Saponin, for molecular biology, used as non-ionic surfactant
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Formaldehyde in Water