Merck
  • AMPK activation modulates IL-36-induced inflammatory responses by regulating IκBζ expression in the skin.
AMPK activation modulates IL-36-induced inflammatory responses by regulating IκBζ expression in the skin.
British journal of pharmacology (2024-03-27)
Yi-Ting Huang, Ling-Ya Chiu, Po-Hsuan Lu, Pa-Fan Hsiao, Jen-Yu Wang, Ping-Hsun Lu, Nan-Lin Wu
ABSTRACT
The interleukin (IL)-36 pathway is a critical player in the pathogenesis of pustular psoriasis. However, therapies targeting this pathway are limited or unaffordable (e.g. the anti-IL-36 receptor antibody). AMP-activated protein kinase (AMPK), a regulator of cellular energy and metabolism, is known to participate in inflammatory diseases. However, its role in IL-36-induced skin inflammation remains unclear. Therefore, we sought to investigate the role of AMPK signals in regulating IL-36-induced responses in the skin. IL-36-stimulated primary normal human epidermal keratinocytes (NHEKs) and IL-36-injected (intradermally) BALB/c mice served as the cell and animal models, respectively. Additionally, 5-aminoimidazole-4-carboxamide riboside (AICAR) and A769662 served as AMPK activators. AICAR and A769662 significantly suppressed the IL-36-induced IL-8 (CXCL8) and CCL20 production from NHEKs. IL-36-induced IκBζ protein expression was prominently reduced and IKK/IκBα phosphorylation was attenuated by AICAR and A769662. Conversely, AMPKα knockdown increased IκBζ protein expression and IKK/IκBα phosphorylation in IL-36-treated NHEKs. Furthermore, AICAR and A769662 enhanced IL-36-induced-IκBζ protein degradation via the proteasome-dependent but not the lysosome-dependent pathway. Pretreatment of NHEKs with IL-36 slightly suppressed the AICAR- and A769662-triggered phosphorylation of AMPK and acetyl-CoA carboxylase. In the mouse model, topical application of AICAR significantly reduced ear swelling, redness, epidermal thickening, neutrophil infiltration and inflammatory and antimicrobial peptide gene expression. AMPK activation suppresses IL-36-induced IL-8 and CCL20 release by regulating IκBζ expression in keratinocytes and reduces IL-36-induced skin inflammation in mice, suggesting that AMPK activation is a potential strategy for treating patients with IL-36-mediated inflammatory skin disorders.
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Peroxidase Conjugated, H+L, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
AICA-Riboside, AICA-Riboside, CAS 2627-69-2, is a cell-permeable nucleoside compound whose phosphorylated metabolite activates AMPK and acts as a regulator of de novo purine synthesis.
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).