Merck
  • GABAA and GABAB receptor dysregulation in superior frontal cortex of subjects with schizophrenia and bipolar disorder.
GABAA and GABAB receptor dysregulation in superior frontal cortex of subjects with schizophrenia and bipolar disorder.
Synapse (New York, N.Y.) (2017-03-21)
S Hossein Fatemi, Timothy D Folsom, Paul D Thuras
ABSTRACT
Schizophrenia and bipolar disorder are complex psychiatric disorders that affect millions of people worldwide. Evidence from gene association and postmortem studies has identified abnormalities of the gamma-aminobutyric acid (GABA) signaling system in both disorders. Abnormal GABAergic signaling and transmission could contribute to the symptomatology of these disorders, potentially through impaired gamma oscillations which normally occur during cognitive processing. In the current study, we examined the protein expression of 14 GABAA and two GABAB receptor subunits in the superior frontal cortex of subjects with schizophrenia, bipolar disorder, and healthy controls. Analyses of Variance (ANOVAs) identified significant group effects for protein levels for the α1, α6, β1, β3, δ, ɛ, and π GABAA receptor subunits and R1 and R2 GABAB receptor subunits. Follow-up t tests confirmed changes for these subunits in subjects with schizophrenia, subjects with bipolar disorder, or both groups. Alterations in stoichiometry of GABA receptor subunits could result in altered ligand binding, transmission, and pharmacology of GABA receptors in superior frontal cortex. Thus, impaired GABAergic transmission may negatively contribute to symptoms such as anxiety or panic as well as impaired learning and information processing, all of which are disrupted in schizophrenia and bipolar disorder. Taken together, these results provide additional evidence of GABAergic receptor abnormalities in these disorders.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-GABA A Receptor α4 Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GABAA Receptor α1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-GABA A Receptor β2 Antibody, cytoplasmic loop, Chemicon®, from rabbit