Merck
  • Inverse regulation of melanoma growth and migration by Orai1/STIM2-dependent calcium entry.
Inverse regulation of melanoma growth and migration by Orai1/STIM2-dependent calcium entry.
Pigment cell & melanoma research (2014-01-30)
Hedwig Stanisz, Stephanie Saul, Cornelia S L Müller, Reinhard Kappl, Barbara A Niemeyer, Thomas Vogt, Markus Hoth, Alexander Roesch, Ivan Bogeski
ABSTRACT
Spontaneous melanoma phenotype switching is controlled by unknown environmental factors and may determine melanoma outcome and responsiveness to anticancer therapy. We show that Orai1 and STIM2 are highly expressed and control store-operated Ca(2+) entry in human melanoma. Lower extracellular Ca(2+) or silencing of Orai1/STIM2 caused a decrease in intracellular Ca(2+) , which correlated with enhanced proliferation and increased expression of microphthalmia-associated transcription factor, a marker for proliferative melanoma phenotype. In contrast, the invasive and migratory potential of melanoma cells was reduced upon silencing of Orai1 and/or STIM2. Accordingly, markers for a non-proliferative, tumor-maintaining phenotype such as JARID1B and Brn2 decreased. Immunohistochemical staining of primary melanomas and lymph node metastases revealed a heterogeneous distribution of Orai1 and STIM2 with elevated expression in the invasive rim of the tumor. In summary, our results support a dynamic model in which Orai1 and STIM2 inversely control melanoma growth and invasion. Pharmacological tuning of Orai1 and particularly STIM2 might thus prevent metastatic spread and render melanomas more susceptible to conventional therapy.
Sigma-Aldrich
Anti-ORAI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-SOX10 antibody produced in mouse, clone 1E6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-STIM2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-POU3F2 (AB2) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-STIM2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution