Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402361

Sigma-Aldrich

Monoclonal Anti-SOX10 antibody produced in mouse

clone 1E6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DOM, MGC15649, WS2E, WS4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.89 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX10(6663)

General description

Anti-AURA Antibody detects endogenous levels of total AURA protein.
SOX10 (SRY-box 10) is a co-transcription factor that is located on human chromosome 22q13. SOX10 is temporarily expressed in melanoblasts.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional activator after forming a protein complex with other proteins. This protein acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease. (provided by RefSeq)

Immunogen

SOX10 (NP_008872, 336 a.a. ~ 433 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KPPGVALPTVSPPGVDAKAQVKTETAGPQGPPHYTDQPSTSQIAYTSLSLPHYGSAFPSISRPQFDYSDHQPSGPYYGHSGQASGLYSAFSYMGPSQR

Biochem/physiol Actions

SOX10 (SRY-box 10) regulates the expression of ErbB3 in neural crest cells. It is required for differentiation of peripheral glia. SOX10 plays an important role in the progression of melanocytes. Mutations in SOX10 result in Waardenburg syndrome and Hirschsprung disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction among SOX10, PAX3 and MITF, three genes altered in Waardenburg syndrome
Bondurand N, et al.
Human Molecular Genetics, 9(13), 1907-1917 (2000)
Takuya E Kishimoto et al.
Veterinary pathology, 55(5), 634-644 (2018-06-02)
Oligodendroglioma is a common brain tumor in dogs, particularly brachycephalic breeds. Oligodendrocyte precursor cells (OPCs) are suspected to be a possible origin of oligodendroglioma, although it has not been well elucidated. In the present study, 27 cases of canine brain
The transcription factor Sox10 is a key regulator of peripheral glial development
Britsch S, et al.
Genes & Development, 15(1), 66-78 (2001)
The SOX10/Sox10 gene from human and mouse: sequence, expression, and transactivation by the encoded HMG domain transcription factor
Pusch C, et al.
Human Genetics, 103(2), 115-123 (1998)
SOX10-Nano-Lantern Reporter Human iPS Cells; A Versatile Tool for Neural Crest Research
Horikiri T, et al.
PLoS ONE (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service