Merck
  • Acid Sphingomyelinase regulates the localization and trafficking of palmitoylated proteins.
Acid Sphingomyelinase regulates the localization and trafficking of palmitoylated proteins.
Biology open (2019-05-31)
Xiahui Xiong, Chia-Fang Lee, Wenjing Li, Jiekai Yu, Linyu Zhu, Yongsoon Kim, Hui Zhang, Hong Sun
ABSTRACT
In human, loss of Acid Sphingomeylinase (ASM/SMPD1) causes Niemann-Pick Disease, type A. ASM hydrolyzes sphingomyelins to produce ceramides but protein targets of ASM remain largely unclear. Our mass-spectrometry-based proteomic analyses have identified >100 proteins associated with the ASM-dependent, detergent-resistant membrane microdomains (lipid rafts), with >60% of these proteins being palmitoylated, including SNAP23, Src-family kinases Yes and Lyn, and Ras and Rab family small GTPases. Inactivation of ASM abolished the presence of these proteins in the plasma membrane, with many of them trapped in the Golgi. While palmitoylation inhibitors and palmitoylation mutants phenocopied the effects of ASM inactivation, we demonstrated that ASM is required for the transport of palmitoylated proteins, such as SNAP23 and Lyn, from the Golgi to the plasma membrane without affecting palmitoylation directly. Importantly, ASM delivered extracellularly can regulate the trafficking of SNAP23 from the Golgi to the plasma membrane. Our studies suggest that ASM, acting at the plasma membrane to produce ceramides, regulates the localization and trafficking of the palmitoylated proteins.
Sigma-Aldrich
Anti-Ras Antibody, (K-, H-, N-), clone 9A11.2, clone 9A11.2, Upstate®, from mouse
Sigma-Aldrich
2-Bromohexadecanoic acid, ~97%
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens