Skip to Content
Merck
All Photos(1)

Key Documents

238422

Sigma-Aldrich

2-Bromohexadecanoic acid

~97%

Synonym(s):

2-Bromopalmitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH(Br)CO2H
CAS Number:
Molecular Weight:
335.32
Beilstein:
1726517
EC Number:
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

~97%

Quality Level

form

solid

SMILES string

CCCCCCCCCCCCCCC(Br)C(O)=O

InChI

1S/C16H31BrO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(17)16(18)19/h15H,2-14H2,1H3,(H,18,19)

InChI key

DPRAYRYQQAXQPE-UHFFFAOYSA-N

Gene Information

human ... PPARD(5467)

Looking for similar products? Visit Product Comparison Guide

General description

PPARδ (peroxisome proliferator-activated receptor, delta isoform) acts as a transcription factor for gene expression as well as playing a role in lipid metabolism regulation; activity of this receptor is ligand-regulated. 2-Bromohexadecanoic acid is a metabolically stable analoge of the fatty acid palmitic acid that has been shown to be a natural ligand for the PPARδ receptor.[1] 2-Bromohexadecanoic acid has also been used in studies of fatty acid oxidation, palmitoylation, and glucose uptake.

Biochem/physiol Actions

2-Bromohexadecanoic acid is a PPARδ agonist. It has also been shown to inhibit fatty acid oxidation, inhibit DHHC-mediated palmitoylation[2], and promote glucose uptake in rat cardiac cells and the insulin-sensitive murine fibroblast line A31-IS[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel Berger et al.
Annual review of medicine, 53, 409-435 (2002-01-31)
The peroxisome proliferator-activated receptors (PPARs) are a group of three nuclear receptor isoforms, PPAR gamma, PPAR alpha, and PPAR delta, encoded by different genes. PPARs are ligand-regulated transcription factors that control gene expression by binding to specific response elements (PPREs)
Kenji Hayata et al.
Journal of pharmacological sciences, 108(3), 348-354 (2008-11-15)
To obtain compounds that promote glucose uptake in muscle cells, the novel cell lines A31-IS derived from Balb/c 3T3 A31 and C2C12-IS from mouse myoblast C2C12 were established. In both cell lines, glucose consumption was induced by insulin and suppressed
Benjamin C Jennings et al.
Journal of lipid research, 50(2), 233-242 (2008-10-02)
Pharmacologic approaches to studying palmitoylation are limited by the lack of specific inhibitors. Recently, screens have revealed five chemical classes of small molecules that inhibit cellular processes associated with palmitoylation (Ducker, C. E., L. K. Griffel, R. A. Smith, S.
Priyanka Tripathi et al.
Journal of lipid research, 62, 100021-100021 (2021-01-01)
Microtubules are polymers composed of αβ-tubulin subunits that provide structure to cells and play a crucial role in in the development and function of neuronal processes and cilia, microtubule-driven extensions of the plasma membrane that have sensory (primary cilia) or
Abiy M Mohammed et al.
Biochemical pharmacology, 85(1), 109-114 (2012-10-25)
Phagocyte-like NADPH oxidase (Nox2) has been shown to play regulatory roles in the metabolic dysfunction of the islet β-cell under the duress of glucolipotoxic conditions and exposure to proinflammatory cytokines. However, the precise mechanisms underlying Nox2 activation by these stimuli

Questions

  1. Does the solution need to be stored away from light?

    1 answer
    1. There is no indication that this product is light sensitive in solution. It is recommended that solutions be stored in aliquots at - 20 deg C for up to 1 month. Keep containers tightly sealed. Avoid repeated freeze/thaw cycles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service