Merck
  • A Novel Spectral Annotation Strategy Streamlines Reporting of mono-ADP-ribosylated Peptides Derived from Mouse Liver and Spleen in Response to IFN-γ.
A Novel Spectral Annotation Strategy Streamlines Reporting of mono-ADP-ribosylated Peptides Derived from Mouse Liver and Spleen in Response to IFN-γ.
Molecular & cellular proteomics : MCP (2021-10-01)
Shiori Kuraoka, Hideyuki Higashi, Yoshihiro Yanagihara, Abhijeet R Sonawane, Shin Mukai, Andrew K Mlynarchik, Mary C Whelan, Michael O Hottiger, Waqas Nasir, Bernard Delanghe, Masanori Aikawa, Sasha A Singh
ABSTRACT
Mass spectrometry-enabled ADP-ribosylation workflows are developing rapidly, providing researchers a variety of ADP-ribosylome enrichment strategies and mass spectrometric acquisition options. Despite the growth spurt in upstream technologies, systematic ADP-ribosyl (ADPr) peptide mass spectral annotation methods are lacking. HCD-dependent ADP-ribosylome studies are common but the resulting MS2 spectra are complex, owing to a mixture of b/y-ions and the m/p-ion peaks representing one or more dissociation events of the ADPr moiety (m-ion) and peptide (p-ion). In particular, p-ions that dissociate further into one or more fragment ions can dominate HCD spectra but are not recognized by standard spectral annotation workflows. As a result, annotation strategies that are solely reliant upon the b/y-ions result in lower spectral scores that in turn reduce the number of reportable ADPr peptides. To improve the confidence of spectral assignments we implemented an ADPr peptide annotation and scoring strategy. All MS2 spectra are scored for the ADPr m-ions, but once spectra are assigned as an ADPr peptide they are further annotated and scored for the p-ions. We implemented this novel workflow to ADPr peptides enriched from the liver and spleen isolated from mice post 4-hour exposure to systemic IFN-γ. HCD collision energy experiments were first performed on the Orbitrap Fusion Lumos and the Q Exactive, with notable ADPr peptide dissociation properties verified with CID (Lumos). The m-ion and p-ion series score distributions revealed that ADPr peptide dissociation properties vary markedly between instruments and within instrument collision energy settings, with consequences on ADPr peptide reporting and amino acid localization. Consequentially, we increased the number of reportable ADPr peptides by 25% (liver) and 17% (spleen) by validation and the inclusion of lower confidence ADPr peptide spectra. This systematic annotation strategy will streamline future reporting of ADPr peptides that have been sequenced using any HCD/CID-based method.
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
PARP Inhibitor VIII, PJ34, The PARP Inhibitor VIII, PJ34, also referenced under CAS 344458-15-7, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.
Sigma-Aldrich
ADP-HPD, Dihydrate, Ammonium Salt, ADP-HPD, Dihydrate is an amino analog of ADP-ribose that acts as a highly potent, noncompetitive, and specific inhibitor of poly(ADP-ribose) glycohydrolase (PARG; IC₅₀ = 120 nM).
Sigma-Aldrich
2-Chloroacetamide, ≥98%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%