Skip to Content
Merck
All Photos(4)

Key Documents

C0267

Sigma-Aldrich

2-Chloroacetamide

≥98%

Synonym(s):

Chloracetamide, alpha-Chloroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CONH2
CAS Number:
Molecular Weight:
93.51
Beilstein:
878191
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.05 mmHg ( 20 °C)

Quality Level

Assay

≥98%

form

powder

mp

116-118 °C (lit.)

SMILES string

NC(=O)CCl

InChI

1S/C2H4ClNO/c3-1-2(4)5/h1H2,(H2,4,5)

InChI key

VXIVSQZSERGHQP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroacetamidecan be used as a reactant to synthesize:
  • 2-Amino-4-chloroquinolines via o-alkylation of 4-chloro-1H-quinolin-2-ones and subsequent Smiles rearrangement.
  • 2-(Formylaryloxy)acetamides by reacting with hydroxybenzaldehydes in the presence of potassium carbonate as a base.
  • Biarylamine derivatives via aluminum triflate-catalyzed direct amination of benzhydrols.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zhang et al.
Journal of agricultural and food chemistry, 59(9), 4614-4621 (2011-03-23)
A butachlor-degrading strain, designated FLY-8, was isolated from rice field soil and was identified as Paracoccus sp. Strain FLY-8 could degrade and utilize six chloroacetamide herbicides as carbon sources for growth, and the degradation rates followed the order alachlor >
Cleonice Rocha et al.
Environmental pollution (Barking, Essex : 1987), 152(1), 239-244 (2007-07-17)
Solid-phase microextraction coupled with gas chromatography-mass spectrometry (SPME-GC-MS) was used to analyze two triazine (atrazine and simazine) and three chloroacetamide herbicides (acetochlor, alachlor, and metolachlor) in water samples from a midwest US agricultural drainage ditch for two growing seasons. The
Ezgi Odabasi et al.
EMBO reports, 20(6) (2019-04-27)
Centriolar satellites are ubiquitous in vertebrate cells. They have recently emerged as key regulators of centrosome/cilium biogenesis, and their mutations are linked to ciliopathies. However, their precise functions and mechanisms of action remain poorly understood. Here, we generated a kidney
K Eszter Borbas et al.
Bioconjugate chemistry, 18(4), 1205-1212 (2007-06-23)
Targeted radiopharmaceuticals offer the possibility of improved tumor imaging and radiotherapy, with reduced side effects. A variety of monoclonal antibodies and antibody fragments have previously been successfully radiolabeled and used in diagnostic imaging and targeted radiotherapy of cancer. Many such
Michelle L Hladik et al.
Environmental science & technology, 39(17), 6561-6574 (2005-09-30)
Although laboratory studies have revealed that many different neutral degradates of chloroacetamide herbicides can form during thermochemical, biological, and photochemical transformations, relatively few have been sought in the environment, despite their likely generation in appreciable amounts, relative persistence, and known

Articles

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service