Merck
  • An exercise "sweet spot" reverses cognitive deficits of aging by growth-hormone-induced neurogenesis.
An exercise "sweet spot" reverses cognitive deficits of aging by growth-hormone-induced neurogenesis.
iScience (2021-11-12)
Daniel G Blackmore, Frederik J Steyn, Alison Carlisle, Imogen O'Keeffe, King-Year Vien, Xiaoqing Zhou, Odette Leiter, Dhanisha Jhaveri, Jana Vukovic, Michael J Waters, Perry F Bartlett
ABSTRACT
Hippocampal function is critical for spatial and contextual learning, and its decline with age contributes to cognitive impairment. Exercise can improve hippocampal function, however, the amount of exercise and mechanisms mediating improvement remain largely unknown. Here, we show exercise reverses learning deficits in aged (24 months) female mice but only when it occurs for a specific duration, with longer or shorter periods proving ineffective. A spike in the levels of growth hormone (GH) and a corresponding increase in neurogenesis during this sweet spot mediate this effect because blocking GH receptor with a competitive antagonist or depleting newborn neurons abrogates the exercise-induced cognitive improvement. Moreover, raising GH levels with GH-releasing hormone agonist improved cognition in nonrunners. We show that GH stimulates neural precursors directly, indicating the link between raised GH and neurogenesis is the basis for the substantially improved learning in aged animals.
Sigma-Aldrich
Bovine Serum Albumin solution, 7.5% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Donepezil hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Millipore
MILLIPLEX® Mouse High Sensitivity T Cell Panel - Immunology Multiplex Assay, Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
BIM 23056, ≥95% (HPLC), solid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
DAPI, for nucleic acid staining