Merck
3-hydroxy-L-kynurenamine is an immunomodulatory biogenic amine.
Nature communications (2021-07-23)
Cristina C Clement, Angelo D'Alessandro, Sangeetha Thangaswamy, Samantha Chalmers, Raquel Furtado, Sheila Spada, Giada Mondanelli, Federica Ianni, Sarah Gehrke, Marco Gargaro, Giorgia Manni, Luisa Carlota Lopez Cara, Peter Runge, Wanxia Li Tsai, Sinem Karaman, Jorge Arasa, Ruben Fernandez-Rodriguez, Amanda Beck, Antonio Macchiarulo, Massimo Gadina, Cornelia Halin, Francesca Fallarino, Mihaela Skobe, Marc Veldhoen, Simone Moretti, Silvia Formenti, Sandra Demaria, Rajesh K Soni, Roberta Galarini, Roccaldo Sardella, Gregoire Lauvau, Chaim Putterman, Kari Alitalo, Ursula Grohmann, Laura Santambrogio
ABSTRACT
Tryptophan catabolism is a major metabolic pathway utilized by several professional and non-professional antigen presenting cells to maintain immunological tolerance. Here we report that 3-hydroxy-L-kynurenamine (3-HKA) is a biogenic amine produced via an alternative pathway of tryptophan metabolism. In vitro, 3-HKA has an anti-inflammatory profile by inhibiting the IFN-γ mediated STAT1/NF-κΒ pathway in both mouse and human dendritic cells (DCs) with a consequent decrease in the release of pro-inflammatory chemokines and cytokines, most notably TNF, IL-6, and IL12p70. 3-HKA has protective effects in an experimental mouse model of psoriasis by decreasing skin thickness, erythema, scaling and fissuring, reducing TNF, IL-1β, IFN-γ, and IL-17 production, and inhibiting generation of effector CD8+ T cells. Similarly, in a mouse model of nephrotoxic nephritis, besides reducing inflammatory cytokines, 3-HKA improves proteinuria and serum urea nitrogen, overall ameliorating immune-mediated glomerulonephritis and renal dysfunction. Overall, we propose that this biogenic amine is a crucial component of tryptophan-mediated immune tolerance.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, purified by recrystallization
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Citrate Buffer, pH 6.0, 10×, Antigen Retriever, for immunohistochemistry
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g