Merck
  • Cellular stress promotes NOD1/2-dependent inflammation via the endogenous metabolite sphingosine-1-phosphate.
Cellular stress promotes NOD1/2-dependent inflammation via the endogenous metabolite sphingosine-1-phosphate.
The EMBO journal (2021-05-05)
Gang Pei, Joanna Zyla, Lichun He, Pedro Moura-Alves, Heidrun Steinle, Philippe Saikali, Laura Lozza, Natalie Nieuwenhuizen, January Weiner, Hans-Joachim Mollenkopf, Kornelia Ellwanger, Christine Arnold, Mojie Duan, Yulia Dagil, Mikhail Pashenkov, Ivo Gomperts Boneca, Thomas A Kufer, Anca Dorhoi, Stefan He Kaufmann
ABSTRACT
Cellular stress has been associated with inflammation, yet precise underlying mechanisms remain elusive. In this study, various unrelated stress inducers were employed to screen for sensors linking altered cellular homeostasis and inflammation. We identified the intracellular pattern recognition receptors NOD1/2, which sense bacterial peptidoglycans, as general stress sensors detecting perturbations of cellular homeostasis. NOD1/2 activation upon such perturbations required generation of the endogenous metabolite sphingosine-1-phosphate (S1P). Unlike peptidoglycan sensing via the leucine-rich repeats domain, cytosolic S1P directly bound to the nucleotide binding domains of NOD1/2, triggering NF-κB activation and inflammatory responses. In sum, we unveiled a hitherto unknown role of NOD1/2 in surveillance of cellular homeostasis through sensing of the cytosolic metabolite S1P. We propose S1P, an endogenous metabolite, as a novel NOD1/2 activator and NOD1/2 as molecular hubs integrating bacterial and metabolic cues.
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
N-Oleoylethanolamine, ~98% (TLC)
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
Sigma-Aldrich
Saponin, for molecular biology, used as non-ionic surfactant
Sigma-Aldrich
SKI II, ≥98% (HPLC), solid
Sigma-Aldrich
GW4869, ≥90% (NMR)
Sigma-Aldrich
Jasplakinolide, ≥97% (HPLC)
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Vinblastine sulfate salt, ≥97% (HPLC)
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)
Sigma-Aldrich
1-Deoxynojirimycin hydrochloride
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Thapsigargin Ready Made Solution, ≥98% (HPLC), 10 mM in DMSO
Sigma-Aldrich
Ceranib-2, ≥98% (HPLC)
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Actinonin
Sigma-Aldrich
Nigericin sodium salt, ≥98% (TLC)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
L-Cycloserine
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.