Merck
Reprogramming CBX8-PRC1 function with a positive allosteric modulator.
Cell chemical biology (2021-10-30)
Junghyun L Suh, Daniel Bsteh, Bryce Hart, Yibo Si, Tyler M Weaver, Carina Pribitzer, Roy Lau, Shivani Soni, Heather Ogana, Justin M Rectenwald, Jacqueline L Norris, Stephanie H Cholensky, Cari Sagum, Jessica D Umana, Dongxu Li, Brian Hardy, Mark T Bedford, Shannon M Mumenthaler, Heinz-Josef Lenz, Yong-Mi Kim, Gang Greg Wang, Ken H Pearce, Lindsey I James, Dmitri B Kireev, Catherine A Musselman, Stephen V Frye, Oliver Bell
ABSTRACT
Canonical targeting of Polycomb repressive complex 1 (PRC1) to repress developmental genes is mediated by cell-type-specific, paralogous chromobox (CBX) proteins (CBX2, 4, 6, 7, and 8). Based on their central role in silencing and their dysregulation associated with human disease including cancer, CBX proteins are attractive targets for small-molecule chemical probe development. Here, we have used a quantitative and target-specific cellular assay to discover a potent positive allosteric modulator (PAM) of CBX8. The PAM activity of UNC7040 antagonizes H3K27me3 binding by CBX8 while increasing interactions with nucleic acids. We show that treatment with UNC7040 leads to efficient and selective eviction of CBX8-containing PRC1 from chromatin, loss of silencing, and reduced proliferation across different cancer cell lines. Our discovery and characterization of UNC7040 not only reveals the most cellularly potent CBX8-specific chemical probe to date, but also corroborates a mechanism of Polycomb regulation by non-specific CBX nucleotide binding activity.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Cbx7 Antibody, from rabbit, purified by affinity chromatography