Merck
  • ATP13A3 is a major component of the enigmatic mammalian polyamine transport system.
ATP13A3 is a major component of the enigmatic mammalian polyamine transport system.
The Journal of biological chemistry (2020-12-15)
Norin Nabil Hamouda, Chris Van den Haute, Roeland Vanhoutte, Ragna Sannerud, Mujahid Azfar, Rupert Mayer, Álvaro Cortés Calabuig, Johannes V Swinnen, Patrizia Agostinis, Veerle Baekelandt, Wim Annaert, Francis Impens, Steven H L Verhelst, Jan Eggermont, Shaun Martin, Peter Vangheluwe
ABSTRACT
Polyamines, such as putrescine, spermidine, and spermine, are physiologically important polycations, but the transporters responsible for their uptake in mammalian cells remain poorly characterized. Here, we reveal a new component of the mammalian polyamine transport system using CHO-MG cells, a widely used model to study alternative polyamine uptake routes and characterize polyamine transport inhibitors for therapy. CHO-MG cells present polyamine uptake deficiency and resistance to a toxic polyamine biosynthesis inhibitor methylglyoxal bis-(guanylhydrazone) (MGBG), but the molecular defects responsible for these cellular characteristics remain unknown. By genome sequencing of CHO-MG cells, we identified mutations in an unexplored gene, ATP13A3, and found disturbed mRNA and protein expression. ATP13A3 encodes for an orphan P5B-ATPase (ATP13A3), a P-type transport ATPase that represents a candidate polyamine transporter. Interestingly, ATP13A3 complemented the putrescine transport deficiency and MGBG resistance of CHO-MG cells, whereas its knockdown in WT cells induced a CHO-MG phenotype demonstrated as a decrease in putrescine uptake and MGBG sensitivity. Taken together, our findings identify ATP13A3, which has been previously genetically linked with pulmonary arterial hypertension, as a major component of the mammalian polyamine transport system that confers sensitivity to MGBG.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
DL-α-Difluoromethylornithine hydrochloride hydrate, solid, ≥97% (NMR)
Sigma-Aldrich
Penicillin-Streptomycin, Solution Stabilized, with 5,000 units penicillin and 5mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Anti-Early Endosomal Antigen 1 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Millipore
FluorSave Reagent
Sigma-Aldrich
BioTracker Spermine Polyamine Green Fluorescent Live Cell Probe
Sigma-Aldrich
Dynasore hydrate
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ATP13A2 (C-terminal region) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
BioTracker Putrescine Polyamine Green Fluorescent Live Cell Probe
Sigma-Aldrich
BioTracker Spermidine Polyamine Green Fluorescent Live Cell Probe