Skip to Content
Merck
All Photos(1)

Key Documents

345789

Millipore

FluorSave Reagent

Synonym(s):

mounting medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NA.75

form

liquid

Quality Level

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

10-30°C

General description

An aqueous mounting medium for use in conjunction with fluorescent tracers such as fluorescein, rhodamine and phycobiliprotein probes. Specifically designed for preserving fluorescent stained tissue sections. FluorSave is formulated without glycerol and therefore is ideal for use with phycobiliproteins.
Aqueous mounting medium for use in conjunction with fluorescent tracers such as fluorescein, rhodamine, and phycobiliprotein probes. Preserves fluorescence-stained tissue sections. Specially formulated without glycerol. Ideal for use with phycobiliprotein tracers.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sivakumar Vijayaraghavalu et al.
Biomaterials, 227, 119558-119558 (2019-10-28)
Advanced-stage cancers often metastasize to bone, and is the major cause of cancer-related morbidity and mortality. Due to poor biodistribution of intravenously administered anticancer drugs within the bone, chemotherapy is not optimally effective in treating bone metastasis. Additionally, overexpression of
Srinidhi Jayaraman et al.
Journal of neurochemistry, 153(1), 120-137 (2019-09-06)
Previous studies documented up-regulation of peptidase neurolysin (Nln) after brain ischemia, however, the significance of Nln function in the post-stroke brain remained unknown. The aim of this study was to assess the functional role of Nln in the brain after
Sonal Prasad et al.
Communications biology, 3(1), 792-792 (2020-12-29)
The stereocilia of the inner ear sensory cells contain the actin-binding protein radixin, encoded by RDX. Radixin is important for hearing but remains functionally obscure. To determine how radixin influences hearing sensitivity, we used a custom rapid imaging technique to
Yao Yuan et al.
Nature communications, 11(1), 1472-1472 (2020-03-21)
The Hippo TEAD-transcriptional regulators YAP1 and TAZ are central for cell renewal and cancer growth; however, the specific downstream gene networks involved in their activity are not completely understood. Here we introduce TEADi, a genetically encoded inhibitor of the interaction
Frederik Walter et al.
PloS one, 15(9), e0239369-e0239369 (2020-10-01)
Cancer vaccinations sensitize the immune system to recognize tumor-specific antigens de novo or boosting preexisting immune responses. Dendritic cells (DCs) are regarded as the most potent antigen presenting cells (APCs) for induction of (cancer) antigen-specific CD8+ T cell responses. Chitosan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service