Merck
Ligand polarizability contributes to tau fibril binding affinity.
Bioorganic & medicinal chemistry (2011-08-02)
Jordan R Jensen, Katryna Cisek, Nicolette S Honson, Jeff Kuret
ABSTRACT
Whole brain imaging of tau-bearing neurofibrillary lesions has the potential to improve the premortem diagnosis and staging of Alzheimer's disease. Diverse compounds with high affinity for tau aggregates have been reported from high-throughput screens, but the affinity driving features common among them have not been determined. To identify these features, analogs of compounds discovered by high-throughput screening, including phenothiazine, triarylmethine, benzothiazole, and oxindole derivatives, were tested for their ability to displace fluorescent thioflavin dyes from filaments made from recombinant tau protein or authentic paired helical filaments purified from Alzheimer's disease tissue. When representative members of all scaffolds were assayed, the rank order of binding affinity determined for synthetic and authentic filaments correlated strongly, indicating that synthetic filaments have predictive utility for ligand development. Within individual scaffold families, binding affinity was found to correlate with compound polarizability, consistent with a role for dispersion forces in mediating ligand binding. Overall, the data indicate that polarizability is an important commonality among structurally diverse tau binding ligands, and that affinity for tau aggregates can be maximized by integrating formal assessment of this parameter into ligand discovery efforts.
Sigma-Aldrich
p-Rosolic acid, Dye content 84 %
Sigma-Aldrich
Azure B eosinate
Sigma-Aldrich
Azure B, prepared by direct synthesis
Sigma-Aldrich
Pararosaniline hydrochloride, >85.0% (HPLC)
Sigma-Aldrich
Toluidine Blue O, Technical grade
Sigma-Aldrich
Azure B, certified by the Biological Stain Commission
Sigma-Aldrich
Basic Fuchsin, certified by the Biological Stain Commission, Dye content ≥88 %
Sigma-Aldrich
Basic Fuchsin, Dye content >85 %
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
Malachite Green solution, for microscopy
Sigma-Aldrich
Methylene Blue solution, for microscopy
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Gentian violet, for microscopy (Bact., Hist.)
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Azure A eosinate, powder or crystals
Sigma-Aldrich
Azure A chloride, certified by the Biological Stain Commission, Dye content 70 %
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Azure A chloride, Dye content ≥70 %
Sigma-Aldrich
Azure A chloride, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet Solution