Skip to Content
Merck
All Photos(3)

Key Documents

861049

Sigma-Aldrich

Azure A chloride

certified by the Biological Stain Commission, Dye content 70 %

Synonym(s):

Methylene blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
Colour Index Number:
52005
Beilstein:
3922287
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, 70%

color

blue to very dark blue
very dark green

mp

290 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

λmax

633 nm

ε (extinction coefficient)

≥11000 at 242-246 nm in H2O at -1
≥26000 at 287-291 nm in H2O
≥34000 at 620-634 μm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

NC1=CC(SC(C2=N3)=CC(C=C2)=[N+](C)C)=C3C=C1.[Cl-]

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Azure A has been used to determine the amount of charged dextran sulfate in the nanoparticles for the study involving incorporation of heparin-binding proteins into preformed dextran sulfate-chitosan nanoparticles.

Biochem/physiol Actions

The combination of azure A and silver staining is used for the quantification of glycosaminoglycans in agarose gels. It is also used for the staining of chromosomes in the nucleus. Azure A is a main component of the azure B/polychromed methylene blue-eosin stains family, also referred to as the Romanowsky stain. Azure A is a basic cationic dye.

Suitability

Certified for use in Lillie′s modified Nocht′s method and the NCCLS method for blood smears.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

How Romanowsky stains work and why they remain valuable - including a proposed universal Romanowsky staining mechanism and a rational troubleshooting scheme.
Horobin RW
Biotechnic & Histochemistry, 86, 36-51 (2011)
Quantification and characterization of glycosaminoglycans at the nanogram level by a combined azure A-silver staining in agarose gels.
van de Lest CH, et al.
Analytical Biochemistry, 221, 356-361 (1994)
Paula Zaman et al.
International journal of nanomedicine, 11, 6149-6159 (2016-12-07)
Incorporation of proteins into dextran sulfate (DS)-chitosan (CS) nanoparticles (DSCS NPs) is commonly performed using entrapment procedures, in which protein molecules are mixed with DS and CS until particle formation occurs. As DS is an analog of heparin, the authors
Shuhua Bai et al.
Journal of pharmaceutical sciences, 96(8), 2090-2106 (2007-02-09)
This study was designed to test the hypothesis that positively charged dendrimers form a complex with enoxaparin, a low-molecular weight heparin (LMWH), and that the resulting drug-dendrimer complex is effective in preventing deep vein thrombosis after pulmonary administration. Fourier Transform
P Chieco et al.
The Histochemical journal, 25(8), 569-577 (1993-08-01)
Mitotic index is a clinically important parameter in cancer pathology. We developed a staining method using Toluidine Blue to detect efficiently and rapidly mitotic figures in sections of formalin-fixed paraffin-embedded human and rat tissues. Sections were stained at acid pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service