Merck
  • Study on the constituents of Mexican propolis and their cytotoxic activity against PANC-1 human pancreatic cancer cells.
Study on the constituents of Mexican propolis and their cytotoxic activity against PANC-1 human pancreatic cancer cells.
Journal of natural products (2010-03-24)
Feng Li, Suresh Awale, Yasuhiro Tezuka, Hiroyasu Esumi, Shigetoshi Kadota
ABSTRACT
Three new flavonoids, (2R,3R)-3,5-dihydroxy-7-methoxyflavanone 3-(2-methyl)butyrate (1), (7''R)-8-[1-(4'-hydroxy-3'-methoxyphenyl)prop-2-en-1-yl]chrysin (2), and (7''R)-8-[1-(4'-hydroxy-3'-methoxyphenyl)prop-2-en-1-yl]galangin (3), together with 41 known compounds (4-44) were isolated from a methanolic extract of Mexican propolis. Compounds 2 and 3 are unique natural flavones containing a 1-phenylallyl moiety. The in vitro preferential cytotoxicity of all the isolates was evaluated against a PANC-1 human pancreatic cell line. Compound 3 displayed the most potent preferential cytotoxicity (PC(50) 4.6 microM) in the nutrient-deprived medium (NDM) and triggered apoptosis-like morphological changes in PANC-1 cells.
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl cinnamate, ≥98%, stabilized
Sigma-Aldrich
Chrysin, ≥96.5%
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Benzyl cinnamate, 99%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
trans-Cinnamic acid, 97%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%