Merck
  • Correlative light electron ion microscopy reveals in vivo localisation of bedaquiline in Mycobacterium tuberculosis-infected lungs.
Correlative light electron ion microscopy reveals in vivo localisation of bedaquiline in Mycobacterium tuberculosis-infected lungs.
PLoS biology (2021-01-01)
Antony Fearns, Daniel J Greenwood, Angela Rodgers, Haibo Jiang, Maximiliano G Gutierrez
ABSTRACT
Correlative light, electron, and ion microscopy (CLEIM) offers huge potential to track the intracellular fate of antibiotics, with organelle-level resolution. However, a correlative approach that enables subcellular antibiotic visualisation in pathogen-infected tissue is lacking. Here, we developed correlative light, electron, and ion microscopy in tissue (CLEIMiT) and used it to identify the cell type-specific accumulation of an antibiotic in lung lesions of mice infected with Mycobacterium tuberculosis. Using CLEIMiT, we found that the anti-tuberculosis (TB) drug bedaquiline (BDQ) is localised not only in foamy macrophages in the lungs during infection but also accumulate in polymorphonuclear (PMN) cells.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Peel-A-Way embedding molds, Square S-22
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Durcupan ACM, set for 1 L embedding mixture
Sigma-Aldrich
TWEEN® 80, viscous liquid
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Millipore
Middlebrook 7H9 Broth Base, suitable for microbiology, NutriSelect® Plus,
Recommended for use in isolation and cultivation of Mycobacterium species
Millipore
Middlebrook 7H11 Agar Base, suitable for microbiology, NutriSelect® Plus
Sigma-Aldrich
Thiocarbohydrazide, 98%