Merck
  • HIV-1 Vpr-Induced Proinflammatory Response and Apoptosis Are Mediated through the Sur1-Trpm4 Channel in Astrocytes.
HIV-1 Vpr-Induced Proinflammatory Response and Apoptosis Are Mediated through the Sur1-Trpm4 Channel in Astrocytes.
mBio (2020-12-10)
Ge Li, Tapas Makar, Volodymyr Gerzanich, Sudhakar Kalakonda, Svetlana Ivanova, Edna F R Pereira, Sanketh Andharvarapu, Jiantao Zhang, J Marc Simard, Richard Y Zhao
ABSTRACT
Successful treatment of HIV-infected patients with combinational antiretroviral therapies (cART) can now prolong patients' lives to nearly normal life spans. However, the new challenge faced by many of those HIV-infected patients is chronic neuroinflammation and neurotoxicity that often leads to HIV-associated neurocognitive disorders (HAND). However, the mechanism of neuropathogenesis underlying HAND, especially in those who are under cART, is not well understood. HAND is typically characterized by HIV-mediated glial neuroinflammation and neurotoxicity. However, the severity of HAND does not always correlate with HIV-1 viral load but, rather, with the extent of glial activation, suggesting that other HIV-associated factors might contribute to HAND. HIV-1 viral protein R (Vpr) could be one of those viral factors because of its association with neuroinflammation and neurotoxicity. The objective of this study was to delineate the specific roles of HIV-1 infection and Vpr in the activation of neuroinflammation and neurotoxicity, and the possible relationships with the Sur1-Trpm4 channel that contributes to neuroinflammation and neuronal death. Here, we show that HIV-1 expression correlates with activation of proinflammatory markers (TLR4, TNF-α, and NF-κB) and the Sur1-Trpm4 channel in astrocytes of HIV-infected postmortem human and transgenic Tg26 mouse brain tissues. We further show that Vpr alone activates the same set of proinflammatory markers and Sur1 in a glioblastoma SNB19 cell line that is accompanied by apoptosis. The Sur1 inhibitor glibenclamide significantly reduced Vpr-induced apoptosis. Together, our data suggest that HIV-1 Vpr-induced proinflammatory response and apoptosis are mediated at least in part through the Sur1-Trpm4 channel in astrocytes.IMPORTANCE Effective antiretroviral therapies can now prolong patients' lives to nearly normal life span. The current challenge faced by many HIV-infected patients is chronic neuroinflammation and neurotoxicity that contributes to HIV-associated neurocognitive disorders (HAND). We show here that the expression of HIV-1 infection and Vpr correlates with the activation of proinflammatory markers (Toll-like receptor 4 [TLR4], tumor necrosis factor alpha [TNF-α], and NF-κB) and the sulfonylurea receptor 1 (Sur1)-transient receptor potential melastatin 4 (Trpm4) channel in astrocytes of brain tissues. We further show that an FDA-approved Sur1 inhibitory drug called glibenclamide significantly ameliorates apoptotic astrocytic cell death caused by HIV-1 Vpr, which could potentially open the possibility of repurposing glibenclamide for treating HAND.
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Periodic Acid Solution, 1 g/dL in deionized water
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken