Merck
  • Endoderm-Derived Myeloid-like Metaphocytes in Zebrafish Gill Mediate Soluble Antigen-Induced Immunity.
Endoderm-Derived Myeloid-like Metaphocytes in Zebrafish Gill Mediate Soluble Antigen-Induced Immunity.
Cell reports (2020-10-08)
Xi Lin, Qiuxia Zhou, Guanzhen Lin, Changlong Zhao, Zilong Wen
ABSTRACT
Immune cells in the mucosal barriers of vertebrates are highly heterogeneous in their origin and function. This heterogeneity is further exemplified by the recent discovery of ectoderm-derived immune cells-metaphocytes in zebrafish epidermis. Yet, whether non-hematopoiesis-derived immune cells generally exist in barrier tissues remains obscured. Here, we report the identification and characterization of an endoderm-derived immune cell population in the gill and intestine of zebrafish. Transcriptome analysis reveals that the endoderm-derived immune cells are myeloid-like cells with high similarities to the ectoderm-derived metaphocytes in epidermis. Like metaphocytes in epidermis, the endoderm-derived immune cells are non-phagocytic but professional in external soluble antigen uptake. Depletion of the endoderm-derived immune cells in gill hinder the local immune response to external soluble stimulants. This study demonstrates a general existence of non-hematopoiesis-derived immune cells in zebrafish mucosal barriers and challenges the prevalent view that resident immune cells in mucosal barriers arise exclusively from hematopoiesis.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Supelco
Timestrip PLUS 8 °C 48 hours
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
L-15 Medium (Leibovitz), Without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer