Skip to Content
Merck
All Photos(1)

Key Documents

80826

Supelco

Timestrip PLUS 8 °C 48 hours

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

description

Irreversible, single use

quality

Quality Assurance: Produced under ISO9001:2008

shelf life

limited shelf life, expiry date on the label

storage condition

room temperature

parameter

48 h run-out window ((calibrated at +10 °C)

size

40 mm × 19 mm

General description

Timestrip® PLUS indicators record elapsed time at or above a specified threshold temperature in a simple and quick way. They are small, self-adhesive labels that can be attached to or integrated into a product or device.

Visit our Timestrip Web Portal to learn more

Application

Manual activation button on back. The "ON" window turns blue to confirm it is armed. Blue color appears in the viewing window if a breach of the threshold temperature has occurred. Duration of breaches is shown by progress of blue dye along time markers.

Packaging

80826-10EA: 10 labels in sachet
80826-100EA: 100 labels in sachet
80826-500EA: 500 labels in sachet

Components

Non-toxic, polymeric assembly multilayer and aluminum-plastic laminate encapsulating colored liquid chemicals

Specifications

  • Threshold temperature: 8°C/46°F
  • Usage: calibrated to an isothermal temperature environment of +10°C
  • Time Accuracy: ±15% in time under isothermal conditions
  • Temperature Accuracy: ±1 °C/2 °F

Legal Information

Timestrip is a registered trademark of Timestrip Limited
Timestrip Plus is a trademark of Timestrip Limited

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maciej T Luczynski et al.
Biochemical and biophysical research communications, 501(1), 124-130 (2018-05-02)
Discoidin Domain Receptor 2 (DDR2) is a collagen-binding receptor tyrosine kinase that initiates delayed and sustained tyrosine phosphorylation signalling. To understand the molecular basis of this unique phosphorylation profile, here we utilise fluorescence microscopy to map the spatiotemporal localisation of
Sara Cimini et al.
Neurobiology of disease, 89, 101-111 (2016-01-02)
Alzheimer disease (AD) is the most prevalent form of dementia. Loss of hippocampal synapses is the first neurodegenerative event in AD. Synaptic loss has been associated with the accumulation in the brain parenchyma of soluble oligomeric forms of amyloid β
Tim Sen Wang et al.
Developmental cell, 53(6), 627-645 (2020-06-07)
Mitochondrial outer membrane permeabilization (MOMP) is a core event in apoptosis signaling. However, the underlying mechanism of BAX and BAK pore formation remains incompletely understood. We demonstrate that mitochondria are globally and dynamically targeted by endolysosomes (ELs) during MOMP. In
Mariaelena Repici et al.
Molecular neurobiology, 56(1), 61-77 (2018-04-21)
Mutations in the gene encoding DJ-1 are associated with autosomal recessive forms of Parkinson's disease (PD). DJ-1 plays a role in protection from oxidative stress, but how it functions as an "upstream" oxidative stress sensor and whether this relates to
Irene Hernando-Herraez et al.
Nature communications, 10(1), 4361-4361 (2019-09-27)
Age-related tissue alterations have been associated with a decline in stem cell number and function. Although increased cell-to-cell variability in transcription or epigenetic marks has been proposed to be a major hallmark of ageing, little is known about the molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service