Merck
  • Marine natural products from the Turkish sponge Agelas oroides that inhibit the enoyl reductases from Plasmodium falciparum, Mycobacterium tuberculosis and Escherichia coli.
Marine natural products from the Turkish sponge Agelas oroides that inhibit the enoyl reductases from Plasmodium falciparum, Mycobacterium tuberculosis and Escherichia coli.
Bioorganic & medicinal chemistry (2007-09-04)
Deniz Tasdemir, Bülent Topaloglu, Remo Perozzo, Reto Brun, Rosann O'Neill, Néstor M Carballeira, Xujie Zhang, Peter J Tonge, Anthony Linden, Peter Rüedi
ABSTRACT
The type II fatty acid pathway (FAS-II) is a validated target for antimicrobial drug discovery. An activity-guided isolation procedure based on Plasmodium falciparum enoyl-ACP reductase (PfFabI) enzyme inhibition assay on the n-hexane-, the CHCl(3-) and the aq MeOH extracts of the Turkish marine sponge Agelas oroides yielded six pure metabolites [24-ethyl-cholest-5alpha-7-en-3-beta-ol (1), 4,5-dibromopyrrole-2-carboxylic acid methyl ester (2), 4,5-dibromopyrrole-2-carboxylic acid (3), (E)-oroidin (4), 3-amino-1-(2-aminoimidazoyl)-prop-1-ene (5), taurine (6)] and some minor, complex fatty acid mixtures (FAMA-FAMG). FAMA, consisting of a 1:2 mixture of (5Z,9Z)-5,9-tricosadienoic (7) and (5Z,9Z)-5,9-tetracosadienoic (8) acids, and FAMB composed of 8, (5Z,9Z)-5,9-pentacosadienoic (9) and (5Z,9Z)-5,9-hexacosadienoic (10) acids in approximately 3:3:2 ratio were the most active PfFabI inhibitory principles of the hexane extract (IC(50) values 0.35 microg/ml). (E)-Oroidin isolated as free base (4a) was identified as the active component of the CHCl(3) extract. Compound 4a was a more potent PfFabI inhibitor (IC(50) 0.30 microg/ml=0.77 microM) than the (E)-oroidin TFA salt (4b), the active and major component of the aq MeOH extract (IC(50) 5.0 microg/ml). Enzyme kinetic studies showed 4a to be an uncompetitive PfFabI inhibitor (K(i): 0.4+/-0.2 and 0.8+/-0.2 microM with respect to substrate and cofactor). In addition, FAMA and FAMD (mainly consisting of methyl-branched fatty acids) inhibited FabI of Mycobacterium tuberculosis (MtFabI, IC(50)s 9.4 and 8.2 microg/ml, respectively) and Escherichia coli (EcFabI, IC(50)s 0.5 and 0.07 microg/ml, respectively). The majority of the compounds exhibited in vitro antiplasmodial, as well as trypanocidal and leishmanicidal activities without cytotoxicity towards mammalian cells. This study represents the first marine metabolites that inhibit FabI, a clinically relevant enzyme target from the FAS-II pathway of several pathogenic microorganisms.
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Podophyllotoxin
SAFC
Taurine
Sigma-Aldrich
Miltefosine, ≥98% (perchloric acid titration)