Skip to Content
Merck
All Photos(1)

Key Documents

W381306

Sigma-Aldrich

Taurine

≥98%, FG

Synonym(s):

2-Aminoethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SO3H
CAS Number:
Molecular Weight:
125.15
FEMA Number:
3813
Beilstein:
1751215
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.056
NACRES:
NA.21
Organoleptic:
odorless
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

mp

>300 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

NCCS(O)(=O)=O

InChI

1S/C2H7NO3S/c3-1-2-7(4,5)6/h1-3H2,(H,4,5,6)

InChI key

XOAAWQZATWQOTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Taurine is a sulfur-containing amino acid mainly found in animal tissues. In mammals, it is reported to play an important role in the normal development of fetuses and neonates. Taurine is also one of the key ingredients in energy drinks.

Application


  • Efficacy of taurine-enhanced enteral nutrition in improving the outcomes of critically ill patients: A systematic review and meta-analysis.: This meta-analysis evaluates the effectiveness of taurine-enhanced enteral nutrition in critically ill patients. The results indicate that taurine supplementation significantly improves patient outcomes, including reduced inflammation and enhanced recovery rates (Zhang et al., 2024).

  • Taurine reduces the risk for metabolic syndrome: a systematic review and meta-analysis of randomized controlled trials.: This systematic review and meta-analysis demonstrate that taurine supplementation can significantly reduce the risk of metabolic syndrome. The findings support taurine′s potential as a therapeutic agent in managing metabolic health and preventing related diseases (Tzang et al., 2024).

Biochem/physiol Actions

Non-selective endogenous agonist at glycine receptors.
Non-selective endogenous agonist at glycine receptors. Conditionally essential sulfonated amino acid which modulates apoptosis in some cells; functions in many metabolic activities; a product of methionine and cysteine metabolism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safety issues associated with commercially available energy drinks.
Clauson KA, et al.
Journal of the American Pharmaceutical Association, 48(3), e55-e67 (2008)
Taurine supplementation to a low protein diet during foetal and early postnatal life restores a normal proliferation and apoptosis of rat pancreatic islets.
Boujendar S, et al.
Diabetologia, 45(6), 856-866 (2002)
Marlene Wewalka et al.
The Journal of clinical endocrinology and metabolism, 99(4), 1442-1451 (2014-01-18)
Bile acids (BAs) are newly recognized signaling molecules in glucose and energy homeostasis. Differences in BA profiles with type 2 diabetes mellitus (T2D) remain incompletely understood. The objective of the study was to assess serum BA composition in impaired glucose-tolerant
Tsutomu Suzuki et al.
Wiley interdisciplinary reviews. RNA, 2(3), 376-386 (2011-10-01)
Mitochondrial DNA mutations that cause mitochondrial dysfunction are responsible for a wide spectrum of human diseases, referred to as mitochondrial diseases. Pathogenic point mutations are found frequently in genes encoding mitochondrial (mt) tRNAs, indicating that impaired functioning of mutant mt
Felizia K Voss et al.
Science (New York, N.Y.), 344(6184), 634-638 (2014-05-03)
Regulation of cell volume is critical for many cellular and organismal functions, yet the molecular identity of a key player, the volume-regulated anion channel VRAC, has remained unknown. A genome-wide small interfering RNA screen in mammalian cells identified LRRC8A as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service