Merck
Antitubercular constituents from the stem wood of Cinnamomum kotoense.
Journal of natural products (2005-09-27)
Fu-Chi Chen, Chien-Fang Peng, Ian-Lih Tsai, Ih-Sheng Chen
ABSTRACT
Five new compounds, kotolactone A (1), kotolactone B (2), secokotomolide (3), kotodiol (4), and 2-acetyl-5-dodecylfuran (5), and 36 known compounds have been isolated from the stem wood of Cinnamomum kotoense. The structures of these new compounds were determined by means of spectroscopic analysis. The known butanolides, isoobtusilactone A (6) and lincomolide B (7), showed in vitro antitubercular activities with MIC values of 22.48 and 10.16 microM, respectively, against Mycobacterium tuberculosis 90-221387.
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
2-Acetyl-5-methylfuran, ≥98%, FG
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Syringaldehyde, ≥98%, FG
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)