Recommended Products
Assay
≥95.0% (HPLC)
form
solid
mp
112-116 °C (lit.)
functional group
aldehyde
SMILES string
[H]C(=O)c1ccc(O)cc1
InChI
1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H
InChI key
RGHHSNMVTDWUBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-Hydroxybenzaldehyde may be used as a reactant during the synthesis of 4-(4-hydroxyphenyl)-but-3-ene-2-one (a raspberry ketone precursor) and [(pyrrole-2,5-diyl)-co-(benzylidene)]. It may also be used to prepare nonlinear optical quinolinium single crystals (an OHQ–TMS compound) via a condensation reaction with 1,2-dimethylquinolinium 2,4,6-trimethylbenzenesulfonate in methanol.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
345.2 °F
Flash Point(C)
174 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Autotoxic antibiotic production by a marine Chromobacterium.
Marine biology, 27(4), 281-285 (1974)
Polymerization of pyrrole with 4-hydroxybenzaldehyde over Al-MCM-41 mesoporous aluminosilicate materials.
Research on Chemical Intermediates, 41(9), 6485-6496 (2015)
New phenolic N-methylquinolinium single crystals for second-order nonlinear optics.
Opt. Mater., 45, 136-140 (2015)
Secondary amide-based linkers for solid phase organic synthesis.
Tetrahedron Letters, 38(49), 8465-8468 (1997)
Preparation of a raspberry ketone precursor in the presence of rare earth oxide catalysts.
Applied Catalysis A: General, 504, 654-663 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service