Merck
  • Estimation of Dietary Exposure to Contaminants Transferred from the Packaging in Fatty Dry Foods Based on Cereals.
Estimation of Dietary Exposure to Contaminants Transferred from the Packaging in Fatty Dry Foods Based on Cereals.
Foods (Basel, Switzerland) (2020-08-06)
Antía Lestido-Cardama, Ana Rodríguez Bernaldo de Quirós, Juana Bustos, M Luisa Lomo, Perfecto Paseiro Losada, Raquel Sendón
ABSTRACT
Food packaging has received special attention from the food safety standpoint since it could be a potential source of contamination through the migration of chemical substances from the packaging material into food. The assessment of the exposure through the diet to these contaminants from food packaging is necessary. In this work, an estimation of dietary exposure of the young Spanish population (1-17 years) to target chemicals from packaging for fatty dried foods based on cereals was assessed. For this purpose, a gas chromatography coupled to mass spectrometry (GC-MS) method was developed for screening of volatile and semivolatile compounds, potential migrants from the packaging. Then, this technique was used to quantify 8 target analytes, which were previously identified in the packaging (including phthalates, acetyl tributyl citrate (ATBC), butylated hydroxytoluene (BHT) and octocrylene), in composite food samples of fatty cereals prepared according to the consumption data for different age groups. Among the phthalates, exposure to diethyl phthalate (DEP) was the highest for the three groups considered (0.0761-0.545 µg/kg body weight/day), followed by bis(2-ethylhxyl)phathalate (DEHP), while the lowest mean intake was found for di-n-octyl phathalate (DNOP; 0.00463-0.0209 µg/kg body weight/day). The estimated dietary exposures did not exceed for any of the analytes the corresponding established tolerable daily intakes.
Sigma-Aldrich
Hexarelin, ≥90% (HPLC)
Sigma-Aldrich
2,4-Di-tert-butylphenol, 99%
Sigma-Aldrich
ε-Caprolactam, 99%
Sigma-Aldrich
Diisobutyl phthalate, 99%
Sigma-Aldrich
Methyl palmitate, ≥97%
Sigma-Aldrich
2,6-Di-tert-butyl-1,4-benzoquinone, 98%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Glyceryl trioctanoate, ≥95% (GC)