Merck
  • Epigenomic State Transitions Characterize Tumor Progression in Mouse Lung Adenocarcinoma.
Epigenomic State Transitions Characterize Tumor Progression in Mouse Lung Adenocarcinoma.
Cancer cell (2020-07-25)
Lindsay M LaFave, Vinay K Kartha, Sai Ma, Kevin Meli, Isabella Del Priore, Caleb Lareau, Santiago Naranjo, Peter M K Westcott, Fabiana M Duarte, Venkat Sankar, Zachary Chiang, Alison Brack, Travis Law, Haley Hauck, Annalisa Okimoto, Aviv Regev, Jason D Buenrostro, Tyler Jacks
ABSTRACT
Regulatory networks that maintain functional, differentiated cell states are often dysregulated in tumor development. Here, we use single-cell epigenomics to profile chromatin state transitions in a mouse model of lung adenocarcinoma (LUAD). We identify an epigenomic continuum representing loss of cellular identity and progression toward a metastatic state. We define co-accessible regulatory programs and infer key activating and repressive chromatin regulators of these cell states. Among these co-accessibility programs, we identify a pre-metastatic transition, characterized by activation of RUNX transcription factors, which mediates extracellular matrix remodeling to promote metastasis and is predictive of survival across human LUAD patients. Together, these results demonstrate the power of single-cell epigenomics to identify regulatory programs to uncover mechanisms and key biomarkers of tumor progression.
Sigma-Aldrich
Anti-Caveolin-1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-BATF antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
N-(n-Butyl)thiophosphoric triamide, analytical standard
Sigma-Aldrich
ECM Cell Adhesion Array Kit, colorimetric, The ECM Cell Adhesion Array kit is useful for assessing specific cell surface integrins, monitoring in vitro cell differentiation, or screening potential cell adhesion promoters/inhibitors.