Merck
  • NSM00158 Specifically Disrupts the CtBP2-p300 Interaction to Reverse CtBP2-Mediated Transrepression and Prevent the Occurrence of Nonunion.
NSM00158 Specifically Disrupts the CtBP2-p300 Interaction to Reverse CtBP2-Mediated Transrepression and Prevent the Occurrence of Nonunion.
Molecules and cells (2020-05-22)
Xun Chen, Wentao Zhang, Qian Zhang, Tao Song, Zirui Yu, Zhong Li, Ning Duan, Xiaoqian Dang
ABSTRACT
Carboxyl-terminal binding proteins (CtBPs) are transcription regulators that control gene expression in multiple cellular processes. Our recent findings indicated that overexpression of CtBP2 caused the repression of multiple bone development and differentiation genes, resulting in atrophic nonunion. Therefore, disrupting the CtBP2-associated transcriptional complex with small molecules may be an effective strategy to prevent nonunion. In the present study, we developed an in vitro screening system in yeast cells to identify small molecules capable of disrupting the CtBP2-p300 interaction. Herein, we focus our studies on revealing the in vitro and in vivo effects of a small molecule NSM00158, which showed the strongest inhibition of the CtBP2-p300 interaction in vitro. Our results indicated that NSM00158 could specifically disrupt CtBP2 function and cause the disassociation of the CtBP2-p300-Runx2 complex. The impairment of this complex led to failed binding of Runx2 to its downstream targets, causing their upregulation. Using a mouse fracture model, we evaluated the in vivo effect of NSM00158 on preventing nonunion. Consistent with the in vitro results, the NSM00158 treatment resulted in the upregulation of Runx2 downstream targets. Importantly, we found that the administration of NSM00158 could prevent the occurrence of nonunion. Our results suggest that NSM00158 represents a new potential compound to prevent the occurrence of nonunion by disrupting CtBP2 function and impairing the assembly of the CtBP2-p300-Runx2 transcriptional complex.
Sigma-Aldrich
Anti-SPP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-DCX (AB1) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
NSC 95397, ≥97% (HPLC)
Sigma-Aldrich
Anti-Glutathione-S-Transferase (GST)–Peroxidase Conjugate antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Glutathione Sepharose 4B, Cytiva 17-0756-01, pack of 10 mL
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.