Merck
  • Dietary Fibres Differentially Impact on the Production of Phenolic Acids from Rutin in an In Vitro Fermentation Model of the Human Gut Microbiota.
Dietary Fibres Differentially Impact on the Production of Phenolic Acids from Rutin in an In Vitro Fermentation Model of the Human Gut Microbiota.
Nutrients (2020-06-03)
Jaroslav Havlik, Vittoria Marinello, Andrew Gardyne, Min Hou, William Mullen, Douglas J Morrison, Thomas Preston, Emilie Combet, Christine A Edwards
ABSTRACT
Polyphenols are often ingested alongside dietary fibres. They are both catabolised by, and may influence, the intestinal microbiota; yet, interactions between them and the impact on their resultant microbial products are poorly understood. Dietary fibres (inulin, pectin, psyllium, pyrodextrin, wheat bran, cellulose-three doses) were fermented in vitro with human faeces (n = 10) with and without rutin (20 µg/mL), a common dietary flavonol glycoside. Twenty-eight phenolic metabolites and short chain fatty acids (SCFA) were measured over 24 h. Several phenolic metabolites were produced during fibre fermentation, without rutin. With rutin, 3,4-dihydroxyphenylacetic acid (3,4diOHPAA), 3-hydroxyphenylacetic acid (3OHPAA), 3-(3 hydroxyphenyl)propionic acid (3OHPPA) and 3-(3,4-dihydroxyphenyl)propionic acid (3,4diOHPPA; DOPAC) were produced, with 3,4diOHPAA the most abundant, confirmed by fermentation of 13C labelled quercetin. The addition of inulin, wheat bran or pyrodextrin increased 3,4diOHPAA 2 2.5-fold over 24 h (p < 0.05). Rutin affected SCFA production, but this depended on fibre, fibre concentration and timepoint. With inulin, rutin increased pH at 6 h from 4.9 to 5.6 (p = 0.01) but increased propionic, butyric and isovaleric acid (1.9, 1.6 and 5-fold, p < 0.05 at 24 h). Interactions between fibre and phenolics modify production of phenolic acids and SCFA and may be key in enhancing health benefits.
Sigma-Aldrich
3-Hydroxyphenylacetic acid, ≥99%
Sigma-Aldrich
3-(4-Hydroxy-3-methoxyphenyl)propionic acid, ≥96.0% (T)