Merck
  • The Monocyte-Derived Exosomal CLMAT3 Activates the CtBP2-p300-NF-κB Transcriptional Complex to Induce Proinflammatory Cytokines in ALI.
The Monocyte-Derived Exosomal CLMAT3 Activates the CtBP2-p300-NF-κB Transcriptional Complex to Induce Proinflammatory Cytokines in ALI.
Molecular therapy. Nucleic acids (2020-09-01)
Zhi Chen, Wei-Hua Dong, Zhong-Min Qiu, Qiu-Gen Li
ABSTRACT
Monocytes and macrophages are the two major cell types involved in innate immunity. Exosomes act as signaling molecules to regulate cell-to-cell communication by releasing proteins, mRNAs, microRNAs (miRNAs), and long noncoding RNAs (lncRNAs). However, it is still unclear whether monocyte-derived exosomes are involved in the communication between monocytes and macrophages. In this study, we analyzed the differentially expressed lncRNA profiles in monocytes isolated from blood samples of healthy controls and acute lung injury (ALI) patients. We focused our study on investigating the signaling downstream of CLMAT3 (colorectal liver metastasis-associated transcript 3), a lncRNA that regulated proinflammatory cytokine genes. We revealed that CLMAT3 specifically targeted CtBP2 (C-terminal binding protein 2) and repressed its expression. Elevated CtBP2 acted as a coactivator to assemble a transcriptional complex with histone acetyltransferase p300 and NF-κB (nuclear factor κB) subunits. In vitro coculture and in vivo injection of ALI monocyte-derived exosomes increased the production of proinflammatory cytokines. Importantly, the administration of two CtBP2 inhibitors, NSC95397 and MTOB, could significantly reverse CtBP2-mediated transactivation. Collectively, our results support a model in which monocyte-derived exosomal CLMAT3 activates the CtBP2-p300-NF-κB complex to induce proinflammatory cytokines, thus contributing to the pathogenesis of ALI.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Millipore
ANTI-FLAG® M1 Agarose Affinity Gel
Millipore
Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-MYC , (C-terminal) antibody produced in mouse, clone 9E10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
NSC 95397, ≥97% (HPLC)
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Sigma-Aldrich
Anti-NF-κB p65 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-P300 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Pyridoxamine-5′-phosphate, ≥98.0% (HPLC)
Sigma-Aldrich
Imprint® RNA Immunoprecipitation Kit, High-capacity Protein A magnetic beads for successful RNA Immunoprecipitation,suitable for use with mRNA and microRNA
Sigma-Aldrich
Anti-NF-kB p50/105 Antibody, clone 1N19, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells