Merck
  • Assessing heterogeneity among single embryos and single blastomeres using open microfluidic design.
Assessing heterogeneity among single embryos and single blastomeres using open microfluidic design.
Science advances (2020-06-05)
Elisabet Rosàs-Canyelles, Andrew J Modzelewski, Alisha Geldert, Lin He, Amy E Herr
ABSTRACT
The process by which a zygote develops from a single cell into a multicellular organism is poorly understood. Advances are hindered by detection specificity and sensitivity limitations of single-cell protein tools and by challenges in integrating multimodal data. We introduce an open microfluidic tool expressly designed for same-cell phenotypic, protein, and mRNA profiling. We examine difficult-to-study-yet critically important-murine preimplantation embryo stages. In blastomeres dissociated from less well-studied two-cell embryos, we observe no significant GADD45a protein expression heterogeneity, apparent at the four-cell stage. In oocytes, we detect differences in full-length versus truncated DICER-1 mRNA and protein, which are insignificant by the two-cell stage. Single-embryo analyses reveal intraembryonic heterogeneity, differences between embryos of the same fertilization event and between donors, and reductions in the burden of animal sacrifice. Open microfluidic design integrates with existing workflows and opens new avenues for assessing the cellular-to-molecular heterogeneity inherent to preimplantation embryo development.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Acrylamide/Bis-acrylamide, 30% solution, BioReagent, suitable for electrophoresis, 37.5:1
Sigma-Aldrich
Microcapillary tube, calibrated, capacity 10 μL
Sigma-Aldrich
Anti-GAPDH, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
Tyrode′s Solution, Acidic, liquid, sterile-filtered, suitable for mouse embryo cell culture
Aspirator tube assemblies for calibrated microcapillary pipettes, pkg of 5 ea
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Chorionic Gonadotropin, Human Urine, Standard Grade, Chorionic Gonadotropin, Human Urine, Standard Grade, CAS 9002-61-3, is a native glycoprotein hormone that is synthesized by chorionic tissue of the placenta and found in urine during pregnancy.