Merck
Copper-free click chemistry for dynamic in vivo imaging.
Proceedings of the National Academy of Sciences of the United States of America (2007-10-19)
Jeremy M Baskin, Jennifer A Prescher, Scott T Laughlin, Nicholas J Agard, Pamela V Chang, Isaac A Miller, Anderson Lo, Julian A Codelli, Carolyn R Bertozzi
ABSTRACT
Dynamic imaging of proteins in live cells is routinely performed by using genetically encoded reporters, an approach that cannot be extended to other classes of biomolecules such as glycans and lipids. Here, we report a Cu-free variant of click chemistry that can label these biomolecules rapidly and selectively in living systems, overcoming the intrinsic toxicity of the canonical Cu-catalyzed reaction. The critical reagent, a substituted cyclooctyne, possesses ring strain and electron-withdrawing fluorine substituents that together promote the [3 + 2] dipolar cycloaddition with azides installed metabolically into biomolecules. This Cu-free click reaction possesses comparable kinetics to the Cu-catalyzed reaction and proceeds within minutes on live cells with no apparent toxicity. With this technique, we studied the dynamics of glycan trafficking and identified a population of sialoglycoconjugates with unexpectedly rapid internalization kinetics.
Sigma-Aldrich
Dibenzocyclooctyne-sulfo-N-hydroxysuccinimidyl ester, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-maleimide, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-acid, 95%, storage temp.:-20°C
Sigma-Aldrich
Dibenzocyclooctyne-amine, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-N-hydroxysuccinimidyl ester, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-acid, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-Fluor 545, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-maleimide, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-alcohol, for Copper-free Click Chemistry
Sigma-Aldrich
Sulfo-dibenzocyclooctyne-biotin conjugate, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-biotin conjugate, for Copper-free Click Chemistry
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-N-hydroxysuccinimidyl ester, ≥90%