Merck
  • Investigation of cell death mechanism and activity of esculetin-loaded PLGA nanoparticles on insulinoma cells in vitro.
Investigation of cell death mechanism and activity of esculetin-loaded PLGA nanoparticles on insulinoma cells in vitro.
The Journal of pharmacy and pharmacology (2020-01-25)
Ayse Karatug Kacar, Fatemeh Bahadori, Serife Evrim Kepekci Tekkeli, Gulacti Topcu, Sehnaz Bolkent
ABSTRACT
The purpose of this study was to prepare targeted cancer therapy formulation against insulinoma INS-1 cells and to study its effect on cell death with related mechanisms in vitro. Polylactide-co-glycolide (PLGA) nano-micelles were used for preparation of esculetin nano-formulation (nano-esculetin). The cells were treated with nano-esculetin and free esculetin. Apoptotic and necrotic cell death percentages, cell proliferation, ATP and GTP reductions and insulin levels were investigated on insulinoma INS-1 cells for both free and nano-esculetin formulations. About 50 mg of PLGA was able to carry 20 mg esculetin in 20 ml of formulation. The obtained optimized formulation was 150 nm, with 92% encapsulation efficiency and a slow-release behaviour was observed during release studies. Nano-esculetin bearing 25, 50 and 100 μg esculetin and free esculetin in equivalent doses successfully decreased cell viability. The prevailing cell death mechanism was necrosis. Along with cell proliferation, intracellular insulin and the ratio of ATP and GTP were decreased even with 12.5, 25 and 50 μg esculetin bearing nano-formulation and its equivalent free esculetin. The results revealed that esculetin is able to show its anti-tumor afficacy after loading to PLGA nano-micelles and nano-encapsulation intensifies its cytotoxic activity in vitro. Current study shows that esculetin and its nano formulations are promising agents in treatment of insulinoma.
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Roche
Cell Proliferation ELISA, BrdU (colorimetric), sufficient for ≤1,000 tests
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
HEPES, ≥99.5% (titration)