Merck
  • Shedding light on autophagy coordinating with cell wall integrity signaling to govern pathogenicity of Magnaporthe oryzae.
Shedding light on autophagy coordinating with cell wall integrity signaling to govern pathogenicity of Magnaporthe oryzae.
Autophagy (2019-07-18)
Ziyi Yin, Wanzhen Feng, Chen Chen, Jiayun Xu, Ying Li, Lina Yang, Jingzhen Wang, Xinyu Liu, Wenhao Wang, Chuyun Gao, Haifeng Zhang, Xiaobo Zheng, Ping Wang, Zhengguang Zhang
ABSTRACT
Cells are faced with various stresses during their growth and development, and autophagy is a degradative process in which cells can break down their own components to recycle macromolecules and provide energy under these stresses. For pathogenic fungi that utilize cell wall as the first barrier against external stress, the cell wall integrity (CWI) pathway also provides an essential role in responding to these stresses. However, the specific connection between autophagy and CWI remains elusive in either the model fungi including budding yeast Saccharomyces cerevisiae or the rice blast fungus Magnaporthe oryzae. Here, we provided evidence that the endoplasmic reticulum (ER) stress is highly induced during M. oryzae infection and that CWI MAP kinase kinase MoMkk1 (S. cerevisiae Mkk1/2 homolog) was subject to phosphorylation regulation by MoAtg1, the only identified kinase in the core autophagy machinery. We also identified MoMkk1 serine 115 as the MoAtg1-dependent phosphorylation site and this phosphorylation could activate CWI, similar to that by the conserved MAP kinase kinase kinase MoMck1 (S. cerevisiae Bck1 homolog). Together with the first report of MoMkk1 subjects to phosphorylation regulation by MoAtg1, we revealed a new mechanism by which autophagy coordinates with CWI signaling under ER stress, and this MoAtg1-dependent MoMkk1 phosphorylation is essential for the pathogenicity of M. oryzae.Abbreviations: A/Ala: alanine; Atg: autophagy-related; Bck1: bypass of C kinase 1; co-IP: co-immunoprecipitation; CWI: cell wall integrity;DTT: dithiothreitol; ER: endoplasmic reticulum; GFP: green fluorescent protein; Mo: Magnaporthe oryzae; MAPK: mitogen-activated protein kinase; Mkk1: mitogen-activated protein kinase-kinase 1; MS: mass spectrometry; PAS: phagophore assembly site; RFP: red fluorescent protein; RT: room temperature; S/Ser: serine; Slt2: suppressor of the lytic phenotype 2; T/Thr: threonine; UPR: unfolded protein response; Y2H: yeast two-hybrid screen.
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Supelco
Epoxy embedding medium, hardener DDSA, for microscopy
Sigma-Aldrich
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
Pyridoxine, ≥98%
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
Iodoacetamide, BioUltra
Millipore
Immobilon®-PSQ PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.2 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Millipore
Miracloth, Rapid, effective filtration of homogenates
Sigma-Aldrich
BL21(DE3) Chemically Competent Cells, for protein expression
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%